S9379
Selenium dioxide
≥98%, powder
Synonym(s):
NSC 56753, Selenium oxide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
SeO2
CAS Number:
Molecular Weight:
110.96
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
vapor pressure
1 mmHg ( 157 °C)
Assay
≥98%
form
powder
color
white to light pink
mp
315 °C (subl.) (lit.)
SMILES string
O=[Se]=O
InChI
1S/O2Se/c1-3-2
InChI key
JPJALAQPGMAKDF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service