Skip to Content
Merck
All Photos(1)

Key Documents

S9322

Sigma-Aldrich

Monoclonal Anti-α/β-SNAP antibody produced in mouse

~1 mg/mL, clone 16D1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-N-ethylmaleimide-sensitive factor attachment protein, alpha, Anti-SNAPA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

16D1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~36 kDa
antigen ~38 kDa

species reactivity

bovine, human, chicken, rat

concentration

~1 mg/mL

technique(s)

western blot: 1:5,000 using chicken or rat kidney and brian

isotype

IgG2b

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

Immunogen

recombinant human α-SNAP.

Physical form

Solution in 20 mM sodium phosphate, 150 mM sodium chloride, 50% glycerol, and 3 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service