Skip to Content
Merck
All Photos(1)

Key Documents

S9186

Sigma-Aldrich

SIB 1757

≥99% (HPLC), crystalline

Synonym(s):

6-Methyl-2-(phenylazo)-3-pyridinol

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$139.00

$139.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$139.00

About This Item

Empirical Formula (Hill Notation):
C12H11N3O
CAS Number:
Molecular Weight:
213.24
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$139.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Assay

≥99% (HPLC)

form

crystalline

color

red

SMILES string

Cc1ccc(O)c(\N=N\c2ccccc2)n1

InChI

1S/C12H11N3O/c1-9-7-8-11(16)12(13-9)15-14-10-5-3-2-4-6-10/h2-8,16H,1H3/b15-14+

InChI key

LOCPVWIREQIGNQ-CCEZHUSRSA-N

Gene Information

Application

SIB 1757 may be used in mGluR5 metabotropic glutamate receptor-mediated cell signaling studies.

Biochem/physiol Actions

SIB 1757 is a highly selective mGluR5 metabotropic glutamate receptor antagonist. It non-competitively inhibits glutamate-induced increase in Ca2+ at human metabotropic glutamate receptor 1 (hmGluR1).1 Inhibition of mGlu5 receptor renders protection to neurons2 against methamphetamine-induced oxidative damage.3

Features and Benefits

This compound is featured on the Glutamate Receptors (G Protein Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Bruno et al.
Neuropharmacology, 39(12), 2223-2230 (2000-09-07)
We have used potent and selective non-competitive antagonists of metabotropic glutamate receptor subtype 5 (mGlu5) -- 2-methyl-6-phenylethynylpyridine (MPEP), [6-methyl-2-(phenylazo)-3-pyridinol] (SIB-1757) and [(E)-2-methyl-6-(2-phenylethenyl)pyridine] (SIB-1893) - to examine whether endogenous activation of this particular metabotropic glutamate receptor subtype contributes to neuronal degeneration.
M A Varney et al.
The Journal of pharmacology and experimental therapeutics, 290(1), 170-181 (1999-06-25)
Cell lines expressing the human metabotropic glutamate receptor subtype 5a (hmGluR5a) and hmGluR1b were used as targets in an automated high-throughput screening (HTS) system that measures changes in intracellular Ca2+ ([Ca2+]i) using fluorescence detection. This functional screen was used to
Giuseppe Battaglia et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 22(6), 2135-2141 (2002-03-16)
Methamphetamine (MA), a widely used drug of abuse, produces oxidative damage of nigrostriatal dopaminergic terminals. We examined the effect of subtype-selective ligands of metabotropic glutamate (mGlu) receptors on MA neurotoxicity in mice. MA (5 mg/kg, i.p.; injected three times, every

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service