Skip to Content
Merck
All Photos(2)

Documents

S8647

Sigma-Aldrich

Sulfacetamide sodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9N2NaO3S · H2O
CAS Number:
Molecular Weight:
254.24
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

form

powder

color

white to off-white

solubility

H2O: soluble 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycoplasma

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

NC1=CC=C(S(N([Na])C(C)=O)(=O)=O)C=C1.O

InChI

1S/C8H10N2O3S.Na.H2O/c1-6(11)10-14(12,13)8-4-2-7(9)3-5-8;;/h2-5H,9H2,1H3,(H,10,11);;1H2/q;+1;/p-1

InChI key

IHCDKJZZFOUARO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide

Biochem/physiol Actions

Sulfacetamide is a sulfonamide antibiotic that blocks the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase. Sulfacetamide is a competitive inhibitor of bacterial para-aminobenzoic acid (PABA), which is required for bacterial synthesis of folic acid. It is active against Gram positive bacteria, Gram negative bacteria and Chlamydia. Mode of resistance is via the alteration of dihydropteroate synthase or alternative pathway for folic acid synthesis.

Packaging

100G

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Storage class (TRGS 510): Non Combustible Solids

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margareta Ungureanu et al.
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi, 110(2), 456-459 (2007-09-07)
In the present study we emphasized the antituberculosis action of new sulphacetamide derivatives. In order o extend the research for obtaining antituberculosis substances, we decided to study the influence of the introducing of the thiourea and sulphamide groups in the
Ilknur Akyol-Salman et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 28(1), 49-52 (2011-07-15)
This study aimed to compare the efficacy of topical N-acetyl-cysteine (NAC) with a topical steroid-antibiotic combination, betamethasone-sulfacetamide sodium therapy in patients with meibomian gland dysfunction (MGD). Twenty patients with MGD were prospectively randomized and assigned into 2 groups. The patients
M Gooderham
Skin therapy letter, 14(2), 1-3 (2009-03-31)
Many options exist for the treatment of rosacea, including topical and systemic therapies, laser and light-based therapies, and surgical procedures. A classification system for rosacea identifies 4 subtypes (i.e., erythematotelangiectatic, papulopustular, phymatous, and ocular), which may help guide therapeutic decision-making.
Zoe Diana Draelos
Journal of drugs in dermatology : JDD, 9(3), 234-236 (2010-03-18)
Prior to 1962, some of the most versatile drugs in dermatology were approved by the U.S. Food and Drug Administration (FDA) solely on the basis of safety. One of these is the combination 10% sodium sulfacetamide and 5% sulfur. Sodium
James Q Del Rosso
Cutis, 86(5 Suppl), 16-25 (2011-01-15)
Rosacea is a prevalent inflammatory skin disorder that affects approximately 16 million individuals in the United States. Although its exact etiology is unknown, basic science, histologic evidence, and clinical evidence suggest that it is inflammatory in nature. In this 12-week

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service