Skip to Content
Merck
All Photos(1)

Documents

S8256

Sigma-Aldrich

Serum thymic factor

≥95% (HPLC)

Synonym(s):

FTS, pGlu-Ala-Lys-Ser-Gln-Gly-Gly-Ser-Asn

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H54N12O15
CAS Number:
Molecular Weight:
858.85
MDL number:
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

storage temp.

−20°C

SMILES string

C[C@H](NC(=O)[C@@H]1CCC(=O)N1)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCC(N)=O)C(=O)NCC(=O)NCC(=O)N[C@@H](CO)C(=O)N[C@@H](CC(N)=O)C(O)=O


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biochemical characterisation of a serum thymic factor.
J Bach et al.
Nature, 266(5597), 55-57 (1977-03-03)
J M Pleau et al.
The Journal of biological chemistry, 252(22), 8045-8047 (1977-11-25)
The amino acid sequence of a circulating thymic factor (FTS, facteur thymique sérique) isolated from pig serum has been established as less than Glu-Ala-Lys-Ser-Gln-Gly-Gly-Ser-Asn-OH. This sequence was obtained by Edman analysis both on the intact peptide and after tryptic digestion.
R G Strachan et al.
Journal of medicinal chemistry, 22(5), 586-588 (1979-05-01)
Less than Glu-Ala-Lys-Ser-Gln-Gly-Gly-Ser-Asn, a proposed serum thymic factor, has been synthesized. The protected precursor, less than Glu-Ala-Lys(i-Noc)-ser(Bzl)-Gln-Gly-Gly-Ser(Bzl)-Asn, was prepared by a combination of solid phase and solution methods. The benzyl blocking groups were removed by HF and the i-Noc blocking

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service