Skip to Content
Merck
All Photos(1)

Key Documents

S8072

Sigma-Aldrich

Sertindole

≥97.5% (HPLC)

Synonym(s):

1-[2-[4-[5-Chloro-1-(4-fluorophenyl)-1h-indol-3-yl]-1-piperidinyl]ethyl]-2-imidazolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26ClFN4O
CAS Number:
Molecular Weight:
440.94
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97.5% (HPLC)

form

solid

color

off-white

solubility

DMSO: >10 mg/mL
H2O: <2 mg/mL

originator

Abbott

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)-n2cc(C3CCN(CC3)CCN4CCNC4=O)c5cc(Cl)ccc25

InChI

1S/C24H26ClFN4O/c25-18-1-6-23-21(15-18)22(16-30(23)20-4-2-19(26)3-5-20)17-7-10-28(11-8-17)13-14-29-12-9-27-24(29)31/h1-6,15-17H,7-14H2,(H,27,31)

InChI key

GZKLJWGUPQBVJQ-UHFFFAOYSA-N

Gene Information

Application

Sertindole was used to study the role of 5-HT2C receptor in antipsychotic-induced body weight gain in rats.3

Biochem/physiol Actions

Sertindole is a 5-HT2 serotonin and D2 dopamine receptor antagonist and antipsychotic.
Sertindole readily passes the blood-brain barrier and is metabolized into compounds that show greater affinity for 5-HT2 receptors and less for D2 receptors. It is an effective treatment agent for schizophrenia as it improves cognitive impairment.1 Sertindole also acts as antagonist to human cardiac potassium channel, HERG and produces prolonged QT interval.2

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dominique H Holstein et al.
Journal of psychopharmacology (Oxford, England), 25(12), 1600-1613 (2011-09-06)
Sensory gating, indexed by P50 suppression, and sensorimotor gating, indexed by prepulse inhibition (PPI), are impaired in schizophrenia spectrum disorders. There is considerable evidence that schizophrenia patients treated with atypical antipsychotics exhibit relatively less gating deficits than do other patients
D Rampe et al.
The Journal of pharmacology and experimental therapeutics, 286(2), 788-793 (1998-08-08)
Acquired long QT syndrome is a side effect seen with some pharmacological agents, including antipsychotic drugs, and is associated with the development of ventricular arrhythmias. This syndrome is often caused by the blockade of repolarizing potassium channels the human heart.
Agnieszka Nikiforuk et al.
Psychopharmacology, 220(1), 65-74 (2011-09-16)
Prefrontal cortical dysfunctions, including an impaired ability to shift perceptual attentional set, are core features of schizophrenia. Nevertheless, the effectiveness of second-generation antipsychotic drugs in treating specific prefrontal dysfunctions remains equivocal. To model schizophrenia-like cognitive inflexibility in rats, we evaluated
Lowijs Perquin et al.
CNS drugs, 18 Suppl 2, 19-30 (2004-10-06)
Sertindole is a non-sedating atypical antipsychotic agent with high selectivity for dopaminergic neurons in the mesolimbic system. In pivotal clinical trials, sertindole has demonstrated significantly greater efficacy than placebo against both the positive and negative symptoms of schizophrenia. In addition
Tomasz Kos et al.
Psychopharmacology, 214(4), 911-921 (2010-12-17)
Cognitive deficits, including an impaired ability to shift perceptual attentional set, belong to the core features of schizophrenia, are associated with prefrontal cortical dysfunctions, and may involve glutamate NMDA receptors. Although phencyclidine disturbs cognitive flexibility, little is known about the

Articles

We offer many products related to dopamine receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service