Skip to Content
Merck
All Photos(1)

Key Documents

S7201

Sigma-Aldrich

SR-46349 hemifumarate salt

≥98% (HPLC)

Synonym(s):

4-((3Z)-3-(2-Dimethylaminoethyl)oxyimino-3-(2-fluorophenyl)propen-1-yl)phenol hemifumarate salt, Eplivanserin hemifumarate salt, SR-46349B

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$126.00
25 MG
$486.00

$126.00


Estimated to ship onMay 06, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$126.00
25 MG
$486.00

About This Item

Empirical Formula (Hill Notation):
C19H21FN2O2 · 0.5 C4H4O4
CAS Number:
Molecular Weight:
386.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$126.00


Estimated to ship onMay 06, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to off-white

solubility

DMSO: ≥10 mg/mL

originator

Sanofi Aventis

storage temp.

2-8°C

SMILES string

OC(=O)\C=C\C(O)=O.CN(C)CCO\N=C(\C=C\c1ccc(O)cc1)c2ccccc2F.CN(C)CCO\N=C(\C=C\c3ccc(O)cc3)c4ccccc4F

InChI

1S/2C19H21FN2O2.C4H4O4/c2*1-22(2)13-14-24-21-19(17-5-3-4-6-18(17)20)12-9-15-7-10-16(23)11-8-15;5-3(6)1-2-4(7)8/h2*3-12,23H,13-14H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b2*12-9+,21-19-;2-1+

InChI key

RNLKLYQQDLHHBH-ABDBJYMXSA-N

Gene Information

human ... HTR2A(3356)

Application

SR-46349 hemifumarate salt may be used in cell signaling studies.

Biochem/physiol Actions

ASTAR (Antagonist of Serotonin 5HT2A Receptors).
SR-46349 is an antagonist of 5-HT (2A/2C) receptor and increases the release of dopamine by medial prefrontal cortex in rats.1 It increases the signal transduction induced by 5-HT2 receptor by binding with high affinity.2 SR-46349 inhibits the aggregation of rabbit and human platelets in response to serotonin.3

Features and Benefits

This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Durand et al.
Journal of neuroscience research, 67(1), 86-92 (2001-12-26)
The intrastriatal injection of 6-hydroxydopamine (6-OHDA) in newborn rats produces a marked striatal dopamine (DA) depletion, accompanied by a serotonin (5-HT) hyperinnervation and an up-regulation of 5-HT receptors. The aim of the present study was to investigate whether the increase
Agnès Auclair et al.
The European journal of neuroscience, 20(11), 3073-3084 (2004-12-08)
Addictive properties of drugs of misuse are generally considered to be mediated by an increased release of dopamine (DA) in the ventral striatum. However, recent experiments indicated an implication of alpha1b-adrenergic receptors in behavioural responses to psychostimulants and opiates. We
J M Herbert et al.
Thrombosis and haemostasis, 69(3), 262-267 (1993-03-01)
SR 46349 (trans-4-[(3Z)3-(2-dimethylaminoethyl)oxyimino-3(2-fluorophe nyl) propen-1-yl] phenol, hemifumarate) is the first member of a newly-developed 5-HT2 antagonist series. SR 46349 potently inhibited serotonin-induced aggregation of rabbit and human platelets (IC50 = 1 and 3.9 nM respectively) but had no effect on
Kevin T Ball et al.
Psychopharmacology, 181(4), 676-687 (2005-07-08)
Like amphetamine, a locomotor-activating dose of 3,4-methylenedioxymethamphetamine (MDMA) predominantly excites striatal single-unit activity in freely moving rats. Although both D1- and D2-like dopamine (DA) receptors play important roles in this effect, MDMA, unlike amphetamine, strongly increases both DA and serotonin
Małgorzata Filip
Pharmacological reports : PR, 57(1), 35-46 (2005-04-26)
Previous studies have indicated a role of serotonin (5-HT)2 receptors in modulation of the behavioral effects of cocaine. In the present study, the efficacy of SR 46349B (a 5-HT(2A) receptor antagonist) or SDZ SER-082 (a 5-HT(2C) receptor antagonist) in altering

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service