S6576
ChIP Next Gen Seq Sepharose™
Synonym(s):
Staph-Seq
Select a Size
$304.00
Estimated to ship onJune 01, 2025
Select a Size
About This Item
$304.00
Estimated to ship onJune 01, 2025
Recommended Products
Related Categories
General description
Learn more about ChIP kits and protocol.
Legal Information
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Chromatin Immunoprecipitation qPCR for studying gene regulation across conditions.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service