Skip to Content
Merck
All Photos(1)

Key Documents

S5897

Sigma-Aldrich

Sephadex® G-50

BioReagent, DNA grade, for molecular biology, fine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$362.00

$362.00


Estimated to ship onMay 29, 2025



Select a Size

Change View
25 G
$362.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.52

$362.00


Estimated to ship onMay 29, 2025


grade

Molecular Biology
for molecular biology

product line

BioReagent

form

beads

technique(s)

DNA purification: suitable

matrix

cross-linked dextran

swelling

1 g swells to 9-11 mL

bead diameter

20-80 μm

pore size

exclusion limit (20 bp dsDNA (rA)20)

pH

2-10

foreign activity

DNase, RNase, none detected

storage temp.

room temp

SMILES string

O1C(C(C(C(C1CO)O)O)O)OCC2OC(C(C(C2O)O)O)OCC(O)C(O)C(O)C(O)C=O

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sephadex® G-50, Fine is an often-cited gel filtration medium for desalting and buffer exchange of biomolecules > 30,000 MW. Sephadex® is prepared by crosslinking dextran with epichlorohydrin. Sephadex products differ in their degree of cross-linking and hence in their degree of swelling and their molecular fractionation range. “Fine” indicates a small particle size known for excellent efficiency and short diffusion distances.

S5897-100G′s updated product number is GE17-0573-02

Application

Sephadex® G-50 is suitable for spin column purification of DNA (from contaminating small molecules) for use in hybridizations. It was used to purify PCR products prior to DNA sequencing to improve the sequencing quality.[1]

Features and Benefits

  • Excellent recovery
  • Minimal sample dilution
  • Quick and easy to use

Legal Information

Sephadex is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Torben Riehl et al.
PloS one, 7(11), e49354-e49354 (2012-11-13)
The Amundsen Sea, Antarctica, is amongst the most rapidly changing environments of the world. Its benthic inhabitants are barely known and the BIOPEARL 2 project was one of the first to biologically explore this region. Collected during this expedition, Macrostylis
Aron Fazekas et al.
BioTechniques, 48(4), 277-285 (2010-06-24)
Stutter products are a common artifact in the PCR amplification of frequently used genetic markers that contain mononucleotide simple sequence repeats. Despite the importance of accurate determination of nucleotide sequence and allele size, there has been little progress toward decreasing
Mark Del Campo et al.
Journal of molecular biology, 389(4), 674-693 (2009-04-28)
The DEAD-box proteins CYT-19 in Neurospora crassa and Mss116p in Saccharomyces cerevisiae are broadly acting RNA chaperones that function in mitochondria to stimulate group I and group II intron splicing and to activate mRNA translation. Previous studies showed that the
Ryo Takeuchi et al.
Nucleic acids research, 37(3), 877-890 (2008-12-24)
The LAGLIDADG homing endonuclease (LHE) I-AniI has adopted an extremely efficient secondary RNA splicing activity that is beneficial to its host, balanced against inefficient DNA cleavage. A selection experiment identified point mutations in the enzyme that act synergistically to improve
Katharina Besemer et al.
Applied and environmental microbiology, 75(22), 7189-7195 (2009-09-22)
Streams are highly heterogeneous ecosystems, in terms of both geomorphology and hydrodynamics. While flow is recognized to shape the physical architecture of benthic biofilms, we do not yet understand what drives community assembly and biodiversity of benthic biofilms in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service