Skip to Content
Merck
All Photos(1)

Documents

S5441

Sigma-Aldrich

Stresscopin-related peptide amide human trifluoroacetate salt

Synonym(s):

SRP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C205H358N68O57
Molecular Weight:
4687.46
MDL number:
UNSPSC Code:
51111800

biological source

human

Quality Level

mol wt

4687.46 g/mol

packaging

pkg of 500 μg

color

white

solubility

water: soluble

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... UCN2(90226)

Amino Acid Sequence

His-Pro-Gly-Ser-Arg-Ile-Val-Leu-Ser-Leu-Asp-Val-Pro-Ile-Gly-Leu-Leu-Gln-Ile-Leu-Leu-Glu-Gln-Ala-Arg-Ala-Arg-Ala-Ala-Arg-Glu-Gln-Ala-Thr-Thr-Asn-Ala-Arg-Ile-Leu-Ala-Arg-Val-NH2

Biochem/physiol Actions

A specific ligand for the corticotropin-releasing hormone receptor type 2 (CRHR2).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service