Skip to Content
Merck
All Photos(1)

Key Documents

S5326

Sigma-Aldrich

SB-271046A

≥98% (HPLC)

Synonym(s):

5-Chloro-N-(4-methoxy-3-piperazin-1-yl-phenyl)-3-methyl-2-benzo-thiophenesulfonamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$136.00
25 MG
$534.00

$136.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$136.00
25 MG
$534.00

About This Item

Empirical Formula (Hill Notation):
C20H22ClN3O3S2 · HCl
CAS Number:
Molecular Weight:
488.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$136.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: >20 mg/mL

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

Cl.COc1ccc(NS(=O)(=O)c2sc3ccc(Cl)cc3c2C)cc1N4CCNCC4

InChI

1S/C20H22ClN3O3S2.ClH/c1-13-16-11-14(21)3-6-19(16)28-20(13)29(25,26)23-15-4-5-18(27-2)17(12-15)24-9-7-22-8-10-24;/h3-6,11-12,22-23H,7-10H2,1-2H3;1H

InChI key

RMXZRJYGJMSDQK-UHFFFAOYSA-N

Biochem/physiol Actions

SB-271046A is a 5-HT6 receptor antagonist.
The first potent and selective 5-HT6 antagonist. Training in cognitive tasks, as well as administration SB-271046, induces an increase in pERK1/2 and pCREB1 levels, while CREB2 levels are significantly reduced; cognition-enhancing properties of SB-271046 are attributed to the effect on pERK1/2, not pCREB1/2. SB-271046 reverses MK-801-induced, but not scopolamine-induced, learning impairments. However, with coadministration of galanthamine, both types of learning impairments were ameliorated.

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chaofeng Zhu et al.
Frontiers in pharmacology, 11, 475-475 (2020-05-20)
Recent studies have documented that reduced M-current promotes epileptogenesis and attenuates synaptic remodeling. Neurite growth is closely related to the level of 5-HT6 receptor (5-HT6R) in the central nervous system. However, little research is available regarding the relation between 5-HT6R
Anna Cavaccini et al.
Neuron, 98(4), 801-816 (2018-05-01)
Monoaminergic modulation of cortical and thalamic inputs to the dorsal striatum (DS) is crucial for reward-based learning and action control. While dopamine has been extensively investigated in this context, the synaptic effects of serotonin (5-HT) have been largely unexplored. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service