Skip to Content
Merck
All Photos(1)

Key Documents

S5321

Sigma-Aldrich

S15535

≥98% (HPLC), solid

Synonym(s):

1-(2,3-Dihydro-1,4-benzodioxin-5-yl)-4-(2,3-dihydro-1h-inden-2-yl)-piperazine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$695.00

$695.00


Estimated to ship onMay 13, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$695.00

About This Item

Empirical Formula (Hill Notation):
C21H24N2O2
CAS Number:
Molecular Weight:
336.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$695.00


Estimated to ship onMay 13, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: ~6 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

C1COc2c(O1)cccc2N3CCN(CC3)C4Cc5ccccc5C4

InChI

1S/C21H24N2O2/c1-2-5-17-15-18(14-16(17)4-1)22-8-10-23(11-9-22)19-6-3-7-20-21(19)25-13-12-24-20/h1-7,18H,8-15H2

InChI key

QJPPEMXOOWNICQ-UHFFFAOYSA-N

Application

S15535 has been used as a serotonin 1A receptor (5-HT1A) agonist in zebrafish.[1]
S15535 may be used in histamine 5-HT1A-medaited cell signaling studies.

Biochem/physiol Actions

Potent, orally active, partial 5-HT1A receptor agonist
S15535, a benzodioxane, is a selective but partial agonist of histamine serotonin 1A (5-HT1A) receptor. It activates presynaptic 5-HT1A receptors and suppresses the release of hippocampal 5-HT. It acts as an anxiolytic agent and modulates the cholinergic and cognitive function in rodents.[2]

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Dekeyne et al.
Psychopharmacology, 152(1), 55-66 (2000-10-21)
The benzodioxane, S15535, possesses low intrinsic activity and marked selectivity at 5-HT1A receptors, hippocampal populations of which are implicated in anxious states. Herein, we examined its potential anxiolytic actions in relation to its influence upon extracellular levels of 5-HT in
Parallel evaluation of 5-HT1A receptor localization and functionality: autoradiographic studies with [35S]-GTP gamma S and the novel, selective radioligand, [3H]-S 15535.
A Newman-Tancredi et al.
Annals of the New York Academy of Sciences, 861, 263-264 (1999-02-03)
Jasmin M Dao et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 36(7), 1319-1331 (2011-03-18)
Epidemiological studies have shown that adolescent smoking is associated with health risk behaviors, including high-risk sexual activity and illicit drug use. Using rat as an animal model, we evaluated the behavioral and biochemical effects of a 4-day, low-dose nicotine pretreatment
B J van der Vegt et al.
Physiology & behavior, 74(1-2), 205-211 (2001-09-21)
Individual differences in aggressive behaviour have been linked to variability in central serotonergic activity, both in humans and animals. A previous experiment in mice, selectively bred for high or low levels of aggression, showed an up-regulation of postsynaptic serotonin-1A (5-HT(1A))
M J Millan et al.
The Journal of pharmacology and experimental therapeutics, 282(1), 148-161 (1997-07-01)
In these studies, we characterized the influence of the novel benzodioxopiperazine serotonin (5-HT)1A ligand, S 15535, on the release of 5-HT in rat hippocampus and compared its potential anxiolytic properties with those of the 5-HT1A receptor partial agonist, buspirone, the

Articles

We offer many products related to serotonin receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service