Skip to Content
Merck
All Photos(1)

Key Documents

S5071

Sigma-Aldrich

SFK1

≥98% (HPLC), solid

Synonym(s):

Cyclohexylmethyl 4-[imino(octylamino)methyl]benzoate hydrochloride, Suppressor of FK-506 1

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$363.00

$363.00


Estimated to ship onMay 09, 2025



Select a Size

Change View
10 MG
$363.00

About This Item

Empirical Formula (Hill Notation):
C23H37ClN2O2
CAS Number:
Molecular Weight:
409.01
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$363.00


Estimated to ship onMay 09, 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

solubility

DMSO: 12 mg/mL

storage temp.

2-8°C

SMILES string

Cl[H].CCCCCCCCNC(=N)c1ccc(cc1)C(=O)OCC2CCCCC2

InChI

1S/C23H36N2O2.ClH/c1-2-3-4-5-6-10-17-25-22(24)20-13-15-21(16-14-20)23(26)27-18-19-11-8-7-9-12-19;/h13-16,19H,2-12,17-18H2,1H3,(H2,24,25);1H

InChI key

URYYYIJUCLTKBY-UHFFFAOYSA-N

Application

SFK1 may be used in cell signaling studies.

Biochem/physiol Actions

SFK1 interacts directly with yeast mitochodria. Induces cell death in low salt conditions and suppresses the ability of FK506 to inhibit cell growth in the presence of high levels of NaCl.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service