Skip to Content
Merck
All Photos(1)

Key Documents

S4570

Sigma-Aldrich

Sulochrin

≥98% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16O7
CAS Number:
Molecular Weight:
332.30
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

solubility

DMF: soluble
DMSO: soluble
ethanol: soluble
methanol: soluble

antibiotic activity spectrum

fungi

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

COC(=O)c1cc(O)cc(OC)c1C(=O)c2c(O)cc(C)cc2O

InChI

1S/C17H16O7/c1-8-4-11(19)15(12(20)5-8)16(21)14-10(17(22)24-3)6-9(18)7-13(14)23-2/h4-7,18-20H,1-3H3

InChI key

YJRLSCDUYLRBIZ-UHFFFAOYSA-N

Application

Sulochrin was used as standard in LC/ESI for quantification of analytes from moldy food samples.[1]

Biochem/physiol Actions

Antibiotic from Aspergillus and Penicillium sp. Fungal metabolite; VEGF inhibitor and anti-angiogenic that inhibits the VEGF-induced tube formation of human umbilical vein endothelial cells.
Sulochrin, a metabolite of Aspergillus terreus and Penicillium sp., has weak anti-bacterial and anti-fungal properties. It inhibits the degranulation of eosinophils induced by the platelet activating factor.[2][3]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robin D Couch et al.
Journal of biotechnology, 108(2), 171-178 (2004-05-08)
Elimination of undesirable co-metabolites from industrial fermentations is often required due to the toxicities associated with the contaminants and/or due to difficulties in removing the contaminants during downstream processing. Sulochrin is a co-metabolite produced during the Aspergillus terreus lovastatin fermentation.
Sulochrin inhibits eosinophil degranulation.
H Ohashi et al.
The Journal of antibiotics, 50(11), 972-974 (1998-05-21)
H Ohashi et al.
Bioorganic & medicinal chemistry letters, 9(14), 1945-1948 (1999-08-18)
Sulochrin, a metabolite of fungi, has been shown to have an inhibitory activity to eosinophil degranulation. A series of sulochrin derivatives substituted at ortho-positions to the 10-carbonyl group was examined the activity. The importance of alkylester at C-6 position and
J Y Liu et al.
Journal of biotechnology, 114(3), 279-287 (2004-11-04)
Aspergillus fumigatus CY018 was recognized as an endophytic fungus for the first time in the leaf of Cynodon dactylon. By bioassay-guided fractionation, the EtOAc extract of a solid-matrix steady culture of this fungus afforded two new metabolites, named asperfumoid (1)
Michael Sulyok et al.
Analytical and bioanalytical chemistry, 389(5), 1505-1523 (2007-09-18)
This paper describes the extension of a previously published method based on liquid chromatography/electrospray ionization tandem mass spectrometry (HPLC/ESI-MS/MS) from 39 to currently 87 analytes. Besides the mycotoxins for which regulated concentrations exist, the method now comprises not only almost

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service