Skip to Content
Merck
All Photos(1)

Key Documents

S4316

Sigma-Aldrich

Amyloid Precursor Protein β, Secreted human

recombinant, expressed in E. coli (N-terminal histidine tagged), solution

Synonym(s):

Amyloid Precursor Protein β, Secreted, sAPPβ

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$971.00

$971.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$971.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

$971.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

recombinant

expressed in E. coli (N-terminal histidine tagged)

Quality Level

form

solution

mol wt

~100 kDa by SDS-PAGE

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... APP(351)

General description

Amyloid-β (Aβ) peptides are a major component of the senile plaques characteristic of the Alzheimer brain.[1] It is a type- I transmembrane protein. The gene encoding amyloid-β is localized on human chromosome 21.[2][3]

Application

Human amyloid precursor protein β has been used as peptide standard.[4][5]

Biochem/physiol Actions

The secreted amyloid precursor β (sAPPβ) is a proteolytic cleavage product of β amyloid precursor protein (APP). APP is cleaved by β-secretase into two fragments, sAPPβ and β-amyloid peptide (Aβ). The Aβ is a component of amyloid plaques, one of the major hallmarks of Alzheimer`s disease (AD), while the sAPPβ is thought to modulate neuronal function and cell survival. Mutation in APP found in Swedish familial AD pedigree increases the susceptibility of APP to β-secretase cleavage and the formation of Aβ and sAPPβ. In contrast, activation of protein kinase C (PKC) decreases the levels of sAPPβ. The trans-Golgi network (TGN) is the major site for β-secretase activity.

Physical form

0.2 μm filtered solution in 20 mM Tris (pH 7.4), 0.5 M NaCl, 1 mM EDTA, 5 mM βME, 2 μg/ml aprotinin

Preparation Note

sAPPβ was expressed in E. coli as a soluble protein and purified under non-denaturing conditions

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monomeric Amyloid Beta Peptide in Hexafluoroisopropanol Detected by Small Angle Neutron Scattering.
Zhang-Haagen B
PLoS ONE (2016)
The Processing of Amyloid Precursor Protein in the
Central Nervous System of Humans and Rhesus
Macaques
Justyna Anna Dobrowolska
The West Virginia Medical Journal (2013)
D J Selkoe
Nature, 399(6738 Suppl), A23-A31 (1999-07-07)
Studies of the molecular basis of Alzheimer's disease exemplify the increasingly blurred distinction between basic and applied biomedical research. The four genes so far implicated in familial Alzheimer's disease have each been shown to elevate brain levels of the self-aggregating
In vivo BACE1 inhibition leads to brain A? lowering and increased a-secretase processing of APP without effect on Neuregulin-1
Sethu Sankaranarayanan
Journal of Pharmacology and Experimental Therapeutics (2018)
Conformational Dynamics of Specific A? Oligomers Govern Their Ability To Replicate and Induce Neuronal Apoptosis.
Dean DN
Biochemistry (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service