Skip to Content
Merck
All Photos(1)

Documents

S4194

Sigma-Aldrich

SR 12813

≥98%, solid

Synonym(s):

Tetraethyl 2-(3,5-di-tert-butyl-4-hydroxyphenyl)ethenyl-1,1-bisphosphonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H42O7P2
CAS Number:
Molecular Weight:
504.53
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥98%

form

solid

solubility

DMSO: ≥10 mg/mL
H2O: insoluble

originator

GlaxoSmithKline

SMILES string

CCOP(=O)(OCC)C(=C/c1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C)\P(=O)(OCC)OCC

InChI

1S/C24H42O7P2/c1-11-28-32(26,29-12-2)21(33(27,30-13-3)31-14-4)17-18-15-19(23(5,6)7)22(25)20(16-18)24(8,9)10/h15-17,25H,11-14H2,1-10H3

InChI key

YQLJDECYQDRSBI-UHFFFAOYSA-N

Application

SR-12813 was used as control in pregnane X receptor (PXR) binding assay performed in HepG2 liver carcinoma cells and PXR-activation assay mediated by dehydroepiandrosterone in human hepatocytes.

Biochem/physiol Actions

SR-12813 is a 1,1-bisphosphonate ester that exhibits hypocholesterolemic activity. It decreases the biosynthesis of cholesterol by enhancing the degradation of HMG-CoA reductase.
SR 12813 is a pregnane X receptor (PXR, NR 112) agonist; cholesterol lowering drug; HMGCoA reductase inhibitor.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zdeněk Dvořák et al.
EMBO molecular medicine, 12(4), e11621-e11621 (2020-03-11)
The human PXR (pregnane X receptor), a master regulator of drug metabolism, has essential roles in intestinal homeostasis and abrogating inflammation. Existing PXR ligands have substantial off-target toxicity. Based on prior work that established microbial (indole) metabolites as PXR ligands
T A Berkhout et al.
The Journal of biological chemistry, 271(24), 14376-14382 (1996-06-14)
SR-12813 (tetra-ethyl 2-(3,5-di-tert-butyl-4-hydroxyphenyl)ethenyl-1, 1-bisphosphonate) lowers plasma cholesterol in five species. In this paper we investigate the underlying mechanism using Hep G2 cells. SR-12813 inhibited incorporation of tritiated water into cholesterol with an IC50 of 1.2 microM but had no effect
Marija Pinne et al.
PloS one, 11(10), e0164642-e0164642 (2016-10-13)
The pregnane X receptor (PXR/SXR, NR1I2) and constitutive androstane receptor (CAR, NR1I3) are nuclear receptors (NRs) involved in the regulation of many genes including cytochrome P450 enzymes (CYPs) and transporters important in metabolism and uptake of both endogenous substrates and
Krisztina Kohalmy et al.
Drug metabolism and disposition: the biological fate of chemicals, 35(9), 1495-1501 (2007-06-27)
Dehydroepiandrosterone (DHEA), the major precursor of androgens and estrogens, has several beneficial effects on the immune system, on memory function, and in modulating the effects of diabetes, obesity, and chemical carcinogenesis. Treatment of rats with DHEA influences expression of cytochrome
Navdar Sever et al.
The Journal of biological chemistry, 279(41), 43136-43147 (2004-07-13)
Insig-1 and Insig-2 are membrane proteins of the endoplasmic reticulum that regulate lipid metabolism by the following two actions: 1) sterol-induced binding to 3-hydroxy-3-methylglutaryl-coenzyme A reductase, an action that leads to ubiquitination and degradation of the enzyme; and 2) sterol-induced

Articles

Cholesterol synthesis regulation by dietary levels, LDL receptors control lipid-rich LDL particle transport in cells.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service