S2625
Sebacic acid
~95% (GC)
Synonym(s):
Decanedioic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HO2C(CH2)8CO2H
CAS Number:
Molecular Weight:
202.25
Beilstein:
1210591
EC Number:
MDL number:
UNSPSC Code:
12352211
Recommended Products
vapor pressure
1 mmHg ( 183 °C)
Assay
~95% (GC)
bp
294.5 °C/100 mmHg (lit.)
mp
133-137 °C (lit.)
SMILES string
OC(=O)CCCCCCCCC(O)=O
InChI
1S/C10H18O4/c11-9(12)7-5-3-1-2-4-6-8-10(13)14/h1-8H2,(H,11,12)(H,13,14)
InChI key
CXMXRPHRNRROMY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Quality
Titrimetric assay can be misleading because impurities are usually other fatty acids.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service