Skip to Content
Merck
All Photos(1)

Key Documents

S2321

Sigma-Aldrich

Scyllatoxin

recombinant, expressed in E. coli, ≥95% (HPLC), lyophilized powder

Synonym(s):

LeTx I scorpion toxin, Leiurotoxin I

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

recombinant

expressed in E. coli

Assay

≥95% (HPLC)

form

lyophilized powder

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CS)NC(=O)[C@H](CCCCN)NC(=O)CNC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(C)C)NC(=O)CNC(=O)[C@H](CC(C)C)NC(=O)[C@H](CO)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CS)NC(=O)[C@H](CO)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CS)NC(=O)[C@H](CCSC)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CS)NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](C)N)C(=O)NCC(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CS)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CS)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](Cc2c[nH]cn2)C(N)=O

InChI

1S/C142H243N45O39S7/c1-17-75(14)112(139(225)159-59-108(194)163-95(55-110(197)198)130(216)164-80(32-22-25-42-144)121(207)180-98(62-227)135(221)168-85(37-39-109(195)196)123(209)183-102(66-231)137(223)186-111(74(12)13)140(226)170-81(33-23-26-43-145)119(205)171-87(113(149)199)53-78-56-154-68-160-78)187-138(224)103(67-232)182-117(203)79(31-21-24-41-143)161-106(192)57-157-115(201)88(47-69(2)3)173-125(211)90(49-71(6)7)162-107(193)58-158-116(202)89(48-70(4)5)174-131(217)96(60-188)178-120(206)83(35-28-45-156-142(152)153)166-133(219)100(64-229)185-132(218)97(61-189)179-127(213)92(51-73(10)11)175-122(208)84(36-38-104(147)190)167-134(220)99(63-228)181-124(210)86(40-46-233-16)169-118(204)82(34-27-44-155-141(150)151)165-126(212)91(50-72(8)9)176-129(215)94(54-105(148)191)177-136(222)101(65-230)184-128(214)93(172-114(200)76(15)146)52-77-29-19-18-20-30-77/h18-20,29-30,56,68-76,79-103,111-112,188-189,227-232H,17,21-28,31-55,57-67,143-146H2,1-16H3,(H2,147,190)(H2,148,191)(H2,149,199)(H,154,160)(H,157,201)(H,158,202)(H,159,225)(H,161,192)(H,162,193)(H,163,194)(H,164,216)(H,165,212)(H,166,219)(H,167,220)(H,168,221)(H,169,204)(H,170,226)(H,171,205)(H,172,200)(H,173,211)(H,174,217)(H,175,208)(H,176,215)(H,177,222)(H,178,206)(H,179,213)(H,180,207)(H,181,210)(H,182,203)(H,183,209)(H,184,214)(H,185,218)(H,186,223)(H,187,224)(H,195,196)(H,197,198)(H4,150,151,155)(H4,152,153,156)/t75-,76-,79-,80-,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,111-,112-/m0/s1

InChI key

MXWDLLUGULWYIQ-BFRWRHKQSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ala-Phe-Cys-Asn-Leu-Arg-Met-Cys-Gln-Leu-Ser-Cys-Arg-Ser-Leu-Gly-Leu-Leu-Gly-Lys-Cys-Ile-Gly-Asp-Lys-Cys-Glu-Cys-Val-Lys-His-NH2

Biochem/physiol Actions

Recombinant peptide of scyllatoxin, a 31 amino acid long toxin, originally isolated from the venom of the scorpion Leiurus quinquestriatus hebraeus; selective blocker of apamin-sensitive small-conductance Ca2+-activated K+ channels (SK channels) with highest affinity towards Kca2.2.
Scyllatoxin was used to study small-conductance Ca2+-activated K+ channel signaling in rat superior cervical ganglion neurons. Myometrial strips from non-pregnant women were treated with Scyllatoxin to study the NO-induced relaxation of endometrium.

Reconstitution

Stock solution of 1 μm can be obtained by adding 0.292 mL of any conventional buffer per μg of protein.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Martins et al.
Journal of molecular biology, 253(4), 590-603 (1995-11-03)
The three-dimensional solution structure of Scyllatoxin (leiurotoxin I) a venom peptide from the scorpion Leiurus quinquestriatus hebraeus was determined at 1 A resolution by homonuclear proton n.m.r. methods at 500 MHz. Data analysis and structure calculation followed conventional protocols inherent
V Calabro et al.
The journal of peptide research : official journal of the American Peptide Society, 50(1), 39-47 (1997-07-01)
Leiurotoxin I is a neurotoxin, blocker of Ca(2+)-activated apamin-sensitive K+ channel, purified from the venom of the scorpion Leiurus quinquestriatus hebraeus. It is a 31-residue polypeptide reticulated by three disulfide bridges, i.e. Cys3-Cys21, Cys8-Cys26 and Cys12-Cys28. To investigate the role
M Asano et al.
The Journal of pharmacology and experimental therapeutics, 275(2), 775-783 (1995-11-01)
The present study examined the possible role of voltage-dependent Ca++ channels (VDCs) and Ca(++)-activated K+ (KCa) channels in the regulation of resting tone of arteries from spontaneously hypertensive rats (SHR) at a prehypertensive stage. Differences in the effects of agents
Tsuyoshi Akiyama et al.
Neurochemistry international, 56(2), 263-269 (2009-11-10)
To elucidate the role of Ca(2+)-activated K(+) (K(Ca)) channels in the presynaptic acetylcholine (ACh) release from splanchnic nerve endings and the postsynaptic catecholamine release from chromaffin cells, we applied microdialysis technique to the left adrenal medulla of anesthetized rats and
J M Sabatier et al.
Biochemistry, 35(33), 10641-10647 (1996-08-20)
Leiurotoxin I (Lei-NH2), a toxin isolated from the venom of the scorpion Leiurus quinquestriatus hebraeus, is a blocker of the apamin-sensitive Ca(2+)-activated K+ channels. It is a 31-residue polypeptide cross-linked by three disulfide bridges which are presumably between Cys3-Cys21, Cys8-Cys26

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service