Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 vial | Available to ship TODAYfromCarlos Spegazzini | $183.00 |
About This Item
NACRES:
NA.47
UNSPSC Code:
12161501
biological source
mouse (CD-1)
Quality Segment
packaging
vial of 1.0 mL
concentration
20 mg/mL protein
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
mouse ... Rps9(76846)
Biochem/physiol Actions
The hepatic S9 pools from a variety of biological sources represent the post-mitochondrial supernatant fraction from homogenized liver. They are known to be a rich source of drug metabolizing enzymes including P-450. S9 pools are useful in the study of xenobiotic metabolism and drug interactions. They are used in the Ames test, which is a method that uses bacteria to test if a given chemical causes mutations in the genetic material of a given test organism.
Post-mitochondrial supernatant fraction from homogenized liver. Source of drug metabolizing enzymes including P-450, these pools are useful in the study of xenobiotic metabolism and drug interactions.
The hepatic S9 pools from a variety of biological sources represent the post-mitochondrial supernatant fraction from homogenized liver. Known to be a rich source of drug metabolizing enzymes including P-450, these pools are useful in the study of xenobiotic metabolism and drug interactions.
Physical form
Supplied as a solution in 150 mM KCl, 50 mM Tris buffer, pH 7.2 and 2 mM EDTA.
1 of 1
This Item | |||
|---|---|---|---|
| biological source mouse (CD-1) | biological source rat (Sprague-Dawley) | biological source mouse (CD-1) | biological source human |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 100 |
| storage temp. −70°C | storage temp. −70°C | storage temp. −70°C | storage temp. −70°C |
| packaging vial of 1.0 mL | packaging vial of ~20 mg | packaging vial of ~10 mg | packaging vial of ~20 mg |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
| Gene Information mouse ... Rps9(76846) | Gene Information rat ... Mrps9(301371) | Gene Information mouse ... Mgst1(56615) | Gene Information human ... RPS9(6203) |
Still not finding the right product?
Explore all of our products under S9 from Liver, Pooled
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



