Skip to Content
Merck

Skip To

S1951

α-2,3-Sialyltransferase from Pasteurella multocida

recombinant, expressed in E. coli BL21, ≥2 units/mg protein

Synonym(s):

CMP-N-acetylneuraminate:β-D-galactoside α-(2,3)-N-acetylneuraminyltransferase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 unit
Check Cart for Availability
$393.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
MDL number:
Specific activity:
≥2 units/mg protein
Recombinant:
expressed in E. coli BL21

$393.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli BL21

Quality Segment

form

lyophilized powder

specific activity

≥2 units/mg protein

mol wt

46.4 kDa

shipped in

dry ice

storage temp.

−20°C

General description

Sialyltransferases belongs to the glycosyltransferases family with the capability of catalyzing the transfer of N-acetylneuraminic acid residues.[1] It is a multifunctional enzyme with α-2,3-sialyltransferase activity, α-2,6-sialyltransferase activity, sialidase activity, and trans-sialidase activity.[2] It has a molar mass of 46.4 kDa, with pI, pH being 5.94 and 7.5-8.5 respectively.

Biochem/physiol Actions

Sialyltransferase catalyzes the transfer of Neu5Ac from CMP-Neu5Ac as a donor substrate to the β-D-galactosyl-1,4-N-acetyl-D-glucosaminyl termini of acceptor molecules including glycoproteins, glycolipids, and oligosaccharides.[1]
Sialyltransferase transfers Neu5Ac from CMP-Neu5Ac to the galactosyl terminus of acceptor molecules including glycoproteins, glycolipids, and oligosaccharides.

Physical form

Lyophilized powder containing Tris-HCl and NaCl

Preparation Note

Reconstitute the lyophilized powder with a volume of water in the range of 0.1 mL to 1 mL, to give a concentration in the range of 1 unit/mL (1 mL volume of water) to 10 units/mL (0.1 mL volume of water).

Analysis Note

Enzymatic activity assays performed in Tris-HCl buffer (100 mM, pH 8.0) containing CMP-Neu-5Ac (1 mM) and Lac-β−OMU (1 mM) at 37°C for 30 min and analyzed using HPLC with a fluorescence detector (excitation at 325 nm and emission at 372 nm).

Other Notes

One unit will catalyze the formation of 1.0 μmol Neu-5-Ac-α-2,3LacMU from CMP-Neu-5-Ac and Lac-β−OMU per minute at 37 °C at pH 8.0.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S2076S1826C1999
specific activity

≥2 units/mg protein

specific activity

≥5 units/mg protein

specific activity

≥3.0 units/mg protein

specific activity

≥10 units/mg protein

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized solid

recombinant

expressed in E. coli BL21

recombinant

expressed in E. coli BL21

recombinant

expressed in E. coli BL21

recombinant

expressed in E. coli BL21

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

mol wt

46.4 kDa

mol wt

56.8 kDa

mol wt

33.4 kDa

mol wt

26.0 kDa

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters