Skip to Content
Merck
All Photos(1)

Documents

S1940

Sigma-Aldrich

Anti-phospho-Src (pTyr418) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-pp60src(pY418)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

chicken, mouse, human, rat

technique(s)

western blot: 1:1,000 using human platelets or mouse NIH3T3 cells

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr418)

Gene Information

human ... SRC(6714)
mouse ... Src(20779)
rat ... Src(83805)

General description

Src is a proto-oncogene that regulates neuronal functions, such as nerve cell differentiation. The Src kinase family structurally comprises of SH2, SH3 and kinase domains, as well as a negative regulatory C-terminal tail. Src has also been implicated in functions such as cell growth, development and cell-cell interactions . Anti-phospho-Src p(Tyr418) antibody is specific for human, mouse, rat and chicken Src (60 kDa).

Immunogen

The sequence is conserved in human, mouse, chicken and frog.
synthetic phosphopeptide derived from the region of src that contains tyrosine 418, based on the sequence of human src.

Application

Anti-phospho-Src p(Tyr418) antibody is suitable for use in western blot (1:1,000 using human platelets or mouse NIH3T3 cells). The product may also be used for immunoprecipitation (1:1000 using HEK293 or HEL cell lysates ).

Physical form

Solution in Dulbecco′s phosphate-buffered saline (without Mg2+ and Ca2+), pH 7.3, with 50% glycerol, 1.0 mg/ml BSA (IgG and protease free) and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C A Ingraham et al.
Molecular and chemical neuropathology, 10(1), 1-14 (1989-02-01)
The proto-oncogene c-src has been implicated in the development and mature function of the nervous system. pp60c-src, the protein product of the c-src gene, is a tyrosine protein kinase that is highly enriched in fetal neural tissue. pp60c-src appears in
Xiaoming Fan et al.
International journal of molecular sciences, 19(7) (2018-07-25)
Non-coding RNAs are important regulators of protein-coding genes. The current study characterized an antisense long non-coding RNA, ATP1A1-AS1, which is located on the opposite strand of the Na/K-ATPase α1 gene. Our results show that four splice variants are expressed in
María Macarena Massip-Copiz et al.
Archives of biochemistry and biophysics, 616, 1-12 (2017-01-16)
Cystic Fibrosis (CF) is a disease caused by mutations in the cystic fibrosis transmembrane conductance regulator (CFTR) gene. Previously, we found several genes showing a differential expression in CFDE cells (epithelial cells derived from a CF patient). One corresponded to
G Superti-Furga et al.
BioEssays : news and reviews in molecular, cellular and developmental biology, 17(4), 321-330 (1995-04-01)
There is increasing evidence to suggest that cytoplasmic tyrosine kinases of the Src family have a pivotal role in the regulation of a number of cellular processes. Members of this family have been implicated in cellular responses to a variety
J R Engen et al.
Cellular and molecular life sciences : CMLS, 65(19), 3058-3073 (2008-06-20)
Src-family kinases are modular signaling proteins involved in a diverse array of cellular processes. All members of the Src family share the same domain organization, with modular SH3, SH2 and kinase domains followed by a C-terminal negative regulatory tail. X-ray

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service