Skip to Content
Merck
All Photos(3)

Documents

S1450

Sigma-Aldrich

Anti-Superoxide Dismutase (MnSOD) (DD-17)−Cy3 antibody produced in rabbit

1-2 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-IPO-B, Anti-Indophenoloxidase B, Anti-MNSOD, Anti-Manganese Superoxide Dismutase 2, Anti-Manganese dependent SOD, Anti-Mitochondrial SOD, Anti-SOD2, Anti-Superoxide dismutase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

conjugate

CY3 conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

packaging

antibody small pack of 25 μL

concentration

1-2 mg/mL

technique(s)

direct immunofluorescence: 2-5 μg/mL using human HeLa, rat NRK and mouse 3T3 cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOD2(6648)

General description

The SOD2 (superoxide dismutase 2) gene is mapped to human chromosome 6q25.3. The encoded protein functions as a tetramer and is localized to mitochondria.

Immunogen

synthetic peptide corresponding to amino acid residues 183-199 of human superoxide dismutase (MnSOD), conjugated to KLH. The corresponding sequence is conserved in many eukaryotes.

Application

Anti-Superoxide Dismutase (MnSOD) (DD-17)-Cy3 antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

Superoxide dismutase 2 (SOD2) is an antioxidant enzyme and catalyzes the conversion of superoxide to hydrogen peroxide and molecular oxygen. The enzyme offers protection against oxidative damage induced by reactive oxygen species and is also radioprotective. The gene is associated with the metastasis and invasion process of salivary adenoid cystic carcinoma. Manganese SOD is involved in detoxification.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick D Bhola et al.
The Journal of clinical investigation, 126(10), 3827-3836 (2016-09-07)
Upfront resistance to chemotherapy and relapse following remission are critical problems in leukemia that are generally attributed to subpopulations of chemoresistant tumor cells. There are, however, limited means for prospectively identifying these subpopulations, which hinders an understanding of therapeutic resistance.
SOD2 deregulation enhances migration, invasion and has poor prognosis in salivary adenoid cystic carcinoma.
Chang B, et al.
Scientific Reports, 6, 25918-25918 (2016)
Radiation-induced SOD2 overexpression sensitizes colorectal cancer to radiation while protecting normal tissue.
Zhang Z, et al.
Oncotarget, 8(5), 7791-7791 (2017)
Two functional variants of the superoxide dismutase genes in Finnish families with asthma.
Kinnula V L, et al.
Thorax, 59(2), 116-119 (2004)
Substrate-analog binding and electrostatic surfaces of human manganese superoxide dismutase.
Azadmanesh J, et al.
Journal of Structural Biology, 199(1), 68-75 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service