Skip to Content
Merck
All Photos(1)

Key Documents

S1318

Sigma-Aldrich

STO-609-acetic acid

≥95% (HPLC), solid

Synonym(s):

7-Oxo-7H-benzimidazo[2,1-a]benz[de]isoquinoline-3-carboxylic acid - acetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$256.00
25 MG
$807.00

$256.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$256.00
25 MG
$807.00

About This Item

Empirical Formula (Hill Notation):
C19H10N2O3 · C2H4O2
CAS Number:
Molecular Weight:
374.35
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

$256.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

≥95% (HPLC)

form

solid

mp

>300 °C

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC(O)=O.OC(=O)c1ccc2-c3nc4ccccc4n3C(=O)c5cccc1c25

InChI

1S/C19H10N2O3.C2H4O2/c22-18-13-5-3-4-10-11(19(23)24)8-9-12(16(10)13)17-20-14-6-1-2-7-15(14)21(17)18;1-2(3)4/h1-9H,(H,23,24);1H3,(H,3,4)

InChI key

WNRSTFUVBWNELX-UHFFFAOYSA-N

Related Categories

General description

1:1 adduct with acetic acid
STO-609 reduces tumorigenicity of liver cancer cells in vivo.[1] TO-609 reduces the activation of AMP (adenosine monophosphate)-activated protein kinase (AMPK) by ionomycin in a dose dependant manner.[2]

Application

STO609 has been used as a calcium/calmodulin-dependent protein kinase kinase (CAMKK) inhibitor in mouse MLL-AF9 acute myeloid leukemia (AML) cells.[3]

Biochem/physiol Actions

Selective Ca2+/Calmodulin-dependent protein kinase kinase (CaM-KK) antagonist.
Selective Ca2+/Calmodulin-dependent protein kinase kinase (CaM-KK) antagonist. Inhibits CamKKa and CaMKKb with Ki = 80 and 15 ng/mL, respectively. Does not inhibit downstream CaM kinases (CaMKI and CaMKIV).

Features and Benefits

This compound is featured on the Ca/CaMKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The camKK2/camKIV relay is an essential regulator of hepatic cancer
Lin F, et al.
Hepatology, 62(2), 505-520 (2015)
CAMKs support development of acute myeloid leukemia
Kang X, et al.
Journal of Hematology & Oncology, 11(1), 30-30 (2018)
Ca2+/calmodulin-dependent protein kinase kinase-beta acts upstream of AMP-activated protein kinase in mammalian cells
Woods A, et al.
Cell Metabolism, 2(1), 21-33 (2005)
Liping Shen et al.
Toxicology, 421, 22-29 (2019-04-04)
Voltage-gated sodium channels (VGSCs) represent molecular targets for a number of potent neurotoxins that affect the ion permeation or gating kinetics. BmK NT1, an α-scorpion toxin purified from Buthus martensii Karch (BMK), induces excitatory neurotoxicity by activation of VGSCs with
Weiwei Shi et al.
Molecular and cellular biochemistry, 449(1-2), 219-226 (2018-04-21)
Autosomal dominant polycystic kidney disease (ADPKD) is a common heritable human disease. Recently, the role of repressed autophagy in ADPKD has drawn increasing attention. Here, we investigate the mechanism underlying the effect of Saikosaponin-d (SSd), a sarcoplasmic/endoplasmic reticulum Ca2+ ATPase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service