This product has been developed and tested for use with Horseradish Peroxidase antibody conjugates in applications such as ELISA, Western Blot, ImmunoBlot, and Flow Cytometry. This stabilizer has not been tested for use in other applications or with other enzymes. The suitability would have to be determined experimentally by the end user.
S1075
StabilZyme® HRP Conjugate Stabilizer
Synonym(s):
Horseradish Peroxidase Conjugate Stabilizer
Select a Size
$576.00
Select a Size
About This Item
$576.00
Recommended Products
biological source
bovine
Quality Level
form
solution
technique(s)
flow cytometry: suitable
storage temp.
2-8°C
General description
Application
Biochem/physiol Actions
Features and Benefits
- it can be readily incorporated into almost all assay procedures
- conjugates can be added directly to the stabilizer solution, and are then ready for storage and use
- is also an excellent diluent, and often decreases background and increases signal for improved signal-to-noise ratios
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Hello, is this stabilizer can be used for glucose oxidase, lactate oxidate and cholesterol oxidate???
1 answer-
Helpful?
-
-
S1075 StabilZyme® HRP共役安定化剤の比重のデータがあれば、ご教示ください。
1 answer-
The specific gravity is not determined.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service