Skip to Content
Merck
All Photos(1)

Key Documents

S1014

Sigma-Aldrich

Streptonigrin

from Streptomyces flocculus, ≥98% (HPLC), powder, apoptosis inducer

Synonym(s):

Bruneomycin, Nigrin

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$150.00
5 MG
$542.00

$150.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$150.00
5 MG
$542.00

About This Item

Empirical Formula (Hill Notation):
C25H22N4O8
CAS Number:
Molecular Weight:
506.46
Beilstein:
599390
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$150.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Product Name

Streptonigrin from Streptomyces flocculus, ≥98%

Quality Level

Assay

≥98%

antibiotic activity spectrum

viruses

Mode of action

DNA synthesis | interferes

storage temp.

2-8°C

SMILES string

COc1ccc(c(O)c1OC)-c2c(C)c(nc(c2N)-c3ccc4C(=O)C(OC)=C(N)C(=O)c4n3)C(O)=O

InChI

1S/C25H22N4O8/c1-9-14(10-6-8-13(35-2)23(36-3)20(10)30)15(26)19(29-17(9)25(33)34)12-7-5-11-18(28-12)22(32)16(27)24(37-4)21(11)31/h5-8,30H,26-27H2,1-4H3,(H,33,34)

InChI key

PVYJZLYGTZKPJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Streptonigrin (SN) is an aminoquinone antitumour antibiotic. Its antineoplastic activity requires reductive activation by Xanthine-converting enzymes. It induces apoptosis by a mechanism involving NF-κB. DNA cleavage reaction and chromosome damage by SN are influenced by the nature of the metal ion present and dependent on the production of free radicals. Its antibiotic activity is iron-activated.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mi Huang et al.
Veterinary research, 52(1), 48-48 (2021-03-21)
Iron is essential for most bacteria to survive, but excessive iron leads to damage by the Fenton reaction. Therefore, the concentration of intracellular free iron must be strictly controlled in bacteria. Riemerella anatipestifer (R. anatipestifer), a Gram-negative bacterium, encodes the
Laura C C Cook et al.
mBio, 10(3) (2019-06-27)
Streptococcus pyogenes (group A streptococcus [GAS]) is a serious human pathogen with the ability to colonize mucosal surfaces such as the nasopharynx and vaginal tract, often leading to infections such as pharyngitis and vulvovaginitis. We present genome-wide transcriptome sequencing (RNASeq)
Nilanjana Chatterjee et al.
Journal of bacteriology, 202(14) (2020-05-13)
Group A streptococcus (GAS) produces millions of infections worldwide, including mild mucosal infections, postinfection sequelae, and life-threatening invasive diseases. During infection, GAS readily acquires nutritional iron from host heme and hemoproteins. Here, we identified a new heme importer, named SiaFGH
Elaine R Frawley et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(29), 12054-12059 (2013-07-04)
The efficacy of antibiotics and host defenses has been linked to the metabolic and redox states of bacteria. In this study we report that a stress-induced export pump belonging to the major facilitator superfamily effluxes citrate and iron from the
Bryan Knuckley et al.
Chemical communications (Cambridge, England), 46(38), 7175-7177 (2010-08-27)
Protein Arginine Deiminase (PAD) activity is dysregulated in numerous diseases, e.g., Rheumatoid Arthritis. Herein we describe the development of a fluorescence polarization-Activity Based Protein Profiling (fluopol-ABPP) based high throughput screening assay that can be used to identify PAD-selective inhibitors. Using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service