Skip to Content
Merck
All Photos(1)

Key Documents

S0946

Sigma-Aldrich

SB-668875

≥95% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C120H206N44O35S
CAS Number:
Molecular Weight:
2857.26
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

[H]N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CO)C(=O)NCC(=O)N1CCC[C@H]1C(=O)N2CCC[C@H]2C(=O)NCC(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)NCC(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](C)C(=O)N[C@@H](CO)C(=O)NCC(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](Cc3cnc[nH]3)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)NCC(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C(C)O)C(=O)N[C@@H](CCSC)C(N)=O

InChI

1S/C120H206N44O35S/c1-17-60(10)93(115(197)158-77(44-59(8)9)113(195)162-94(65(15)167)116(198)150-68(95(126)177)34-40-200-16)161-91(175)51-137-96(178)61(11)143-97(179)62(12)146-108(190)78(45-66-47-133-55-142-66)157-112(194)79(46-87(125)171)149-89(173)49-139-102(184)80(53-165)159-99(181)64(14)145-105(187)72(29-32-85(123)169)154-110(192)75(42-57(4)5)156-111(193)76(43-58(6)7)155-106(188)70(25-20-37-136-120(131)132)152-107(189)73(30-33-86(124)170)151-98(180)63(13)144-104(186)69(24-19-36-135-119(129)130)147-88(172)48-138-101(183)71(28-31-84(122)168)153-109(191)74(41-56(2)3)148-90(174)50-140-114(196)82-26-21-39-164(82)117(199)83-27-22-38-163(83)92(176)52-141-103(185)81(54-166)160-100(182)67(121)23-18-35-134-118(127)128/h47,55-65,67-83,93-94,165-167H,17-46,48-54,121H2,1-16H3,(H2,122,168)(H2,123,169)(H2,124,170)(H2,125,171)(H2,126,177)(H,133,142)(H,137,178)(H,138,183)(H,139,184)(H,140,196)(H,141,185)(H,143,179)(H,144,186)(H,145,187)(H,146,190)(H,147,172)(H,148,174)(H,149,173)(H,150,198)(H,151,180)(H,152,189)(H,153,191)(H,154,192)(H,155,188)(H,156,193)(H,157,194)(H,158,197)(H,159,181)(H,160,182)(H,161,175)(H,162,195)(H4,127,128,134)(H4,129,130,135)(H4,131,132,136)/t60-,61-,62-,63-,64-,65+,67-,68-,69-,70-,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,93-,94-/m0/s1

InChI key

SURUUKNICBOUOQ-JNZZMZGCSA-N

Amino Acid Sequence

Arg-Ser-Gly-Pro-Pro-Gly-Leu-Gln-Gly-Arg-Ala-Gln-Arg-Leu-Leu-Gln-Ala-Ser-Gly-Asn-His-Ala-Ala-Gly-Ile-Leu-Thr-Met-NH2

Biochem/physiol Actions

SB-668875 is a novel, selective peptide agonist at OX2 orexin receptors. SB-668875 possesses advantages over Orexins A and B which are equipotent at OX2 orexin receptors. The orexins can cause direct and reversible depolarisation of the postsynaptic membrane. The rank order of potencies of orexin receptor agonists to excite serotonergic DRN neurones is orexin-A = orexin-B > SB-668875-DM. In contrast, the rank order of potency of these agonists to excite noradrenergic locus coreleus (LC) neurones is orexin-A > orexin-B > SB-668875-DM.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ellen M Soffin et al.
Neuropharmacology, 46(8), 1168-1176 (2004-04-28)
Electrophysiological recordings from dorsal raphe nucleus (DRN) neurones in rat brain slices have revealed that the orexins can cause direct and reversible depolarisation of the postsynaptic membrane. Whilst it is known that the membrane depolarisation produced by orexin-A can dramatically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service