Skip to Content
Merck
All Photos(1)

Documents

S0885

Sigma-Aldrich

Somatostatin

γ-irradiated, powder, BioXtra, suitable for cell culture

Synonym(s):

Growth hormone release inhibiting factor, SRIF, Somatostatin-14, Somatotropin release inhibiting factor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C76H104N18O19S2
CAS Number:
Molecular Weight:
1637.88
Beilstein:
6436064
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

sterility

γ-irradiated

product line

BioXtra

form

powder

potency

0.3-50 ng/mL

concentration

≥50% (HPLC)

technique(s)

cell culture | mammalian: suitable

solubility

cell culture medium: 10 mL/vial

shipped in

ambient

storage temp.

−20°C

SMILES string

[H]N1[C@@H](Cc2ccccc2)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](Cc6ccccc6)C(=O)N[C@@H]([C@@H](C)O)C(=O)NC(CO)C(=O)N[C@@H](CSSC[C@H](NC(=O)CNC(=O)[C@H](C)N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(N)=O)C1=O)C(O)=O

InChI

1S/C76H104N18O19S2/c1-41(79)64(100)82-37-61(99)83-58-39-114-115-40-59(76(112)113)92-72(108)57(38-95)91-75(111)63(43(3)97)94-71(107)54(33-46-23-11-6-12-24-46)90-74(110)62(42(2)96)93-66(102)51(28-16-18-30-78)84-69(105)55(34-47-36-81-49-26-14-13-25-48(47)49)88-68(104)53(32-45-21-9-5-10-22-45)86-67(103)52(31-44-19-7-4-8-20-44)87-70(106)56(35-60(80)98)89-65(101)50(85-73(58)109)27-15-17-29-77/h4-14,19-26,36,41-43,50-59,62-63,81,95-97H,15-18,27-35,37-40,77-79H2,1-3H3,(H2,80,98)(H,82,100)(H,83,99)(H,84,105)(H,85,109)(H,86,103)(H,87,106)(H,88,104)(H,89,101)(H,90,110)(H,91,111)(H,92,108)(H,93,102)(H,94,107)(H,112,113)/t41-,42+,43+,50-,51-,52-,53-,54-,55-,56-,57?,58-,59-,62-,63-/m0/s1

InChI key

NHXLMOGPVYXJNR-FQSIDJEASA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ala-Gly-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Thr-Ser-Cys

Biochem/physiol Actions

Somatostatin regulates endocrine function and cell proliferation via interaction with G-protein-coupled somatostatin receptors. It exists in two active forms produced by alternative cleavage of a single preproprotein (14 aa and 28 aa forms). Somatosatin inhibits the release of growth hormone; thyroid-stimulating hormone and a range of gastrointestinal hormones: Cholecystokinin (CCK); Enteroglucagon; Gastrin; Gastric inhibitory polypeptide (GIP); Motilin; Secretin; Vasoactive intestinal peptide (VIP).

Physical form

powder-0 °C; stock-frozen in working aliquots, avoid repeated freeze/thaw

Reconstitution

To prepare 2 μg/ml stock solution, add 10 ml sterile culture medium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Henrik H Hansen et al.
The Journal of pharmacology and experimental therapeutics, 350(3), 657-664 (2014-07-06)
Type 2 diabetes is characterized by impaired β-cell function associated with progressive reduction of insulin secretion and β-cell mass. Evidently, there is an unmet need for treatments with greater sustainability in β-cell protection and antidiabetic efficacy. Through an insulin and
Yuko Takano et al.
PloS one, 10(7), e0133812-e0133812 (2015-07-21)
Animal models with defective glucagon action show hyperplasia of islet α-cells, however, the regulatory mechanisms underlying the proliferation of islet endocrine cells remain largely to be elucidated. The Gcggfp/gfp mice, which are homozygous for glucagon/green fluorescent protein knock-in allele (GCGKO)
Amelie Soumier et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(9), 2252-2262 (2014-04-03)
Reduced expression of somatostatin (SST) is reported across chronic brain conditions including major depression and normal aging. SST is a signaling neuropeptide and marker of gamma-amino butyric acid (GABA) neurons, which specifically inhibit pyramidal neuron dendrites. Studies in auditory cortex
Zaki Alsahafi et al.
The Journal of physiology, 593(16), 3673-3692 (2015-05-27)
This study investigates the effects on ventilation of an excitatory stimulus delivered in a spatially and temporally precise manner to the inspiratory oscillator, the preBötzinger complex (preBötC). We used an adeno-associated virus expressing channelrhodopsin driven by the synapsin promoter to
Kanetee Busiah et al.
Hormone research in paediatrics, 82(1), 59-64 (2014-06-20)
Activating mutations in the ABCC8 gene encoding the KATP channel subunit SUR1 cause β-cell dysfunction with non-autoimmune diabetes mellitus in neonates or infants. We investigated whether activating ABCC8 mutations affect endocrine pancreas development. We studied a male infant with compound

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service