Skip to Content
Merck
All Photos(5)

Key Documents

RNV100

Sigma-Aldrich

GENELUTE VIRAL RNA MINIPREP KIT

For the purification of Viral RNA

Synonym(s):

For Preparation of Clean Viral RNA, Viral RNA Extraction Kit, Viral RNA Purification Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52
Pricing and availability is not currently available.

Quality Level

shipped in

ambient

storage temp.

15-25°C

General description

The GenElute Viral RNA Miniprep Kit is designed for isolation of viral RNA in a column-based format. A single protocol is provided for isolation of viral RNA from saline, viral transport medium, saliva, cell suspension, and tissue culture medium. This kit does not separate viral from cellular RNA and will co-purify host cellular RNA if it is present. Efficient purification of low concentrations of viral RNA is enhanced by the inclusion of poly(A) carrier RNA. Downstream compatible applications for purified RNA include RT-qPCR and Next Generation Sequencing.

Suitability

Suitable for Virus RNA Purification

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Skin Corr. 1C - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Liver,Heart

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Products and support for mpox research, offering world-class reagents for immunoassay and molecular assay development.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service