REV888
Fire Monkey / Fire Flower High Molecular Weight-DNA Extraction Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
Includes Columns in 2 mL Collection Tubes, Lysis Solution DNA, Binding Solution, Wash Solution, Elution Buffer
packaging
pkg of Kit (50 extractions)
pkg of Sample Kit (10 extractions)
storage temp.
room temp
Related Categories
General description
For the extraction of high molecular weight DNA from animal and bacterial cells, and size selection of extracted DNA from all sample types.
FIRE MONKEY - HMW DNA EXTRACTION
Fire Monkey is a standard spin-column process which extracts HMW-DNA with average strand lengths of 100kb and above from both bacterial and mammalian cells within 1hr.
FIRE FLOWER - DNA SIZE SELECTION KIT
Fire Flower is a standard spin-column process that can size select extracted DNA from all sample sources within 15mins. Fire Flower can increase the overall average strand length by 30%, while depleting DNA fragments up to 30kb. Fire Flower can be used to size-select up to 200 μL of extracted DNA despite the sample being rich in small DNA fragments.
The two major disruptive features of Fire Monkey are that firstly, it generates average DNA fragment lengths of between 100kbp – 130kbp and secondly, it significantly reduces the amount of small DNA fragments shorter than 10kbp in the final product. This combination, where significant amounts of the DNA fragments have intact lengths of over 165,000 base pairs and up to several hundred thousands base pairs in length, combined with the absence of most of the smaller DNA fragments, through its inbuilt size selection capability, massively improves the eventual long-read sequencing of the sample. Fire Monkey improves the N50 measure of sequencing quality by 100% and yet it takes a sixth of the time to do so compared to the main competing NAIP kits.
FIRE MONKEY - HMW DNA EXTRACTION
Fire Monkey is a standard spin-column process which extracts HMW-DNA with average strand lengths of 100kb and above from both bacterial and mammalian cells within 1hr.
FIRE FLOWER - DNA SIZE SELECTION KIT
Fire Flower is a standard spin-column process that can size select extracted DNA from all sample sources within 15mins. Fire Flower can increase the overall average strand length by 30%, while depleting DNA fragments up to 30kb. Fire Flower can be used to size-select up to 200 μL of extracted DNA despite the sample being rich in small DNA fragments.
The two major disruptive features of Fire Monkey are that firstly, it generates average DNA fragment lengths of between 100kbp – 130kbp and secondly, it significantly reduces the amount of small DNA fragments shorter than 10kbp in the final product. This combination, where significant amounts of the DNA fragments have intact lengths of over 165,000 base pairs and up to several hundred thousands base pairs in length, combined with the absence of most of the smaller DNA fragments, through its inbuilt size selection capability, massively improves the eventual long-read sequencing of the sample. Fire Monkey improves the N50 measure of sequencing quality by 100% and yet it takes a sixth of the time to do so compared to the main competing NAIP kits.
Application
Fire Monkey and Illumina reads used for AMR Characterization:
Rasheed F et al
Microorganisms 8 (9) 1336 Emergence of Resistance to Fluoroquinolones and ThirdGeneration Cephalosporins in Salmonella Typhi in Lahore, Pakistan
Fire Monkey and combined Nanopore/Illumina reads used for complete genome assembly of rare food chain pathogen. Public Health England reference lab:
Gao R et al
Microbiology Resource Announcements Volume: 9, Issue: 35. e0028020 Complete Genome Assemb lies of the Rare Salmonella enterica Serovar Adjame Using Nanopore and Illumina Sequence Reads
Fire Monkey and Nanopore reads used to characterize emerging resistance in a foodchain pathogen subpopulation:
Zhang CZ et al
Journal of Antimicrobial Chemotherapy dkaa288 Emergence of ciprofloxacin heteroresistance in foodborne Salmonella enterica serovar Agona.
Fire Monkey and 10xIllumina reads used for animal chromosome haplotyping. DanaFarber Cancer
Institute, Harvard University:
Tourdot RW & Zhang CZ
bioRxiv Determination of complete chromosomal haplotypes by bulk DNA sequencing
Fire Monkey Total Nucleic Acid and PCR analysis of DNA and messenger RNA used to detect targets of
clinical infectious disease samples:
Todd AV & Lima NE
Nucleic Acid Ratio Determination (Patent Application). Pub. No.: US 2020/0199651
Rasheed F et al
Microorganisms 8 (9) 1336 Emergence of Resistance to Fluoroquinolones and ThirdGeneration Cephalosporins in Salmonella Typhi in Lahore, Pakistan
Fire Monkey and combined Nanopore/Illumina reads used for complete genome assembly of rare food chain pathogen. Public Health England reference lab:
Gao R et al
Microbiology Resource Announcements Volume: 9, Issue: 35. e0028020 Complete Genome Assemb lies of the Rare Salmonella enterica Serovar Adjame Using Nanopore and Illumina Sequence Reads
Fire Monkey and Nanopore reads used to characterize emerging resistance in a foodchain pathogen subpopulation:
Zhang CZ et al
Journal of Antimicrobial Chemotherapy dkaa288 Emergence of ciprofloxacin heteroresistance in foodborne Salmonella enterica serovar Agona.
Fire Monkey and 10xIllumina reads used for animal chromosome haplotyping. DanaFarber Cancer
Institute, Harvard University:
Tourdot RW & Zhang CZ
bioRxiv Determination of complete chromosomal haplotypes by bulk DNA sequencing
Fire Monkey Total Nucleic Acid and PCR analysis of DNA and messenger RNA used to detect targets of
clinical infectious disease samples:
Todd AV & Lima NE
Nucleic Acid Ratio Determination (Patent Application). Pub. No.: US 2020/0199651
Features and Benefits
- Protocols are user-friendly and can be high-throughput.
- Only basic laboratory equipment is required
- Extract and purify long-fragments of DNA within one hour
- In-built size-exclusion limits fragments <10kb
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service