Skip to Content
Merck
All Photos(1)

Documents

RES4055F-A7

SAFC

Ferric citrate

Synonym(s):

Iron(III) citrate tribasic monohydrate, Ferric citrate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5FeO7 · H2O
CAS Number:
Molecular Weight:
262.96
Beilstein:
4052503
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

biological source

synthetic

form

powder

impurities

Trace metals; tested

suitability

suitable for manufacturing use

SMILES string

OC12CC(=O)O[Fe](OC(=O)C1)OC2=O

InChI

1S/C6H8O7.Fe/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);/q;+3/p-3

InChI key

NPFOYSMITVOQOS-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.
To request documentation for this product, please contact Customer Support and select ‘Product Documentation′. Please note that access to documentation for this product requires a confidentiality disclosure agreement.

Packaging

Product is available in the following package sizes:
RES4055F-A702X: 1 kg container
RES4055F-A703X: 5 kg container

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Françoise Guéraud et al.
Free radical biology & medicine, 83, 192-200 (2015-03-07)
The end products of polyunsaturated fatty acid (PUFA) peroxidation, such as malondialdehyde (MDA), 4-hydroxynonenal (HNE), and isoprostanes (8-iso-PGF2α), are widely used as systemic lipid oxidation/oxidative stress biomarkers. However, some of these compounds have also a dietary origin. Thus, replacing dietary
Jimena Carrillo-Tripp et al.
The Journal of general virology, 95(Pt 10), 2285-2296 (2014-07-06)
The cell line IPLB-LD-652Y, derived from the gypsy moth (Lymantria dispar L.), is routinely used to study interactions between viruses and insect hosts. Here we report the full genome sequence and biological characteristics of a small RNA virus, designated Lymantria
Hanh N Lam et al.
PloS one, 9(8), e106115-e106115 (2014-08-30)
The type III secretion system (T3SS) is required for virulence in the gram-negative plant pathogen Pseudomonas syringae pv. tomato DC3000. The alternative sigma factor HrpL directly regulates expression of T3SS genes via a promoter sequence, often designated as the "hrp
Jun Wang et al.
Scientific reports, 5, 10729-10729 (2015-06-04)
Lactoferrin (Lf) can bind to lactoferrin receptor (LfR), leading to iron transport through the plasma membrane. Besides iron transportation, Lf also has antioxidant and anti-inflammatory properties. In the brain, Lf is only synthesized by activated microglia. LfR is present in
Hiroshi Kawabata et al.
Experimental hematology, 43(5), 404-413 (2015-01-31)
Hepcidin is the central regulator of systemic iron homeostasis; dysregulation of hepcidin expression causes various iron metabolic disorders, including hereditary hemochromatosis and anemia of inflammation. To identify molecules that modulate hepcidin expression, we developed a bioassay system for hepcidin gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service