Skip to Content
Merck
All Photos(2)

Key Documents

RES0704P-A7

SAFC

PIPES, disodium salt

Synonym(s):

PIPES disodium salt, 1,4-Piperazinediethanesulfonic acid disodium salt, Piperazine-1,4-bis(2-ethanesulfonic acid) disodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$247.00
1 KG
$879.00
10 KG
$6,149.00

$247.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$247.00
1 KG
$879.00
10 KG
$6,149.00

About This Item

Empirical Formula (Hill Notation):
C8H16N2Na2O6S2
CAS Number:
Molecular Weight:
346.33
Beilstein:
9453872
EC Number:
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.21

$247.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

synthetic

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin and microbial; tested

useful pH range

6.1-7.5

pKa (25 °C)

6.8

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)CCN1CCN(CC1)CCS([O-])(=O)=O

InChI

1S/C8H18N2O6S2.2Na/c11-17(12,13)7-5-9-1-2-10(4-3-9)6-8-18(14,15)16;;/h1-8H2,(H,11,12,13)(H,14,15,16);;/q;2*+1/p-2

InChI key

GMHSTJRPSVFLMT-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Buffer quality is vital for the success of biopharmaceutical processes, because buffers are indispensable in nearly every production step.

Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Application

PIPES Disodium is the sodium salt form of PIPES and is a biological buffer often referred to as a “Good′s” buffer. The pKa of PIPES is 6.8 which makes it ideal for biological applications at physiological conditions. PIPES Disodium is considered to be non-toxic to culture cell lines, highly water soluble and provides high-solution clarity.

PIPES Disodium can be used in a wide array of biological applications including as a component in media formulations. Specific applications include buffers for cell culture and diagnostic reagent production.

Packaging

Product is available in the following package sizes:
RES0704P-A701X: 100 gm container
RES0704P-A702X: 1 kg container
RES0704P-A704X: 10 kg container

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service