Skip to Content
Merck

Skip To

RABBLOCK

5x Blocking Solution (Item F)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 mL
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
$49.20

About This Item

NACRES:
NA.32
UNSPSC Code:
12161703

$49.20

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shipped in

wet ice

storage temp.

−20°C

Other Notes

This item is for RAB series CBEL ELISA kits.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RABFIX1RABCTYROD20-145
storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters