Skip to Content
Merck
All Photos(1)

Key Documents

RAB1021

Sigma-Aldrich

Human Angiotensinogen ELISA Kit

for cell culture supernatants, plasma, and serum samples

Synonym(s):

Angiotensinogen , SERPINA8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84
Pricing and availability is not currently available.

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum sample(s)
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.22 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AGT(183)

General description

This ELISA antibody pair detects Human Angiotensinogen (AGT/SERPINA8)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Janša et al.
Scientific reports, 11(1), 20870-20870 (2021-10-24)
Endometriosis is a common non-malignant gynecological disease that significantly compromises fertility and quality of life of the majority of patients. The gold standard for diagnosis is visual inspection of the pelvic organs by surgical laparoscopy and there are no biomarkers
Ivan L Salazar et al.
Journal of neuroinflammation, 19(1), 44-44 (2022-02-10)
Multiple sclerosis is an inflammatory and degenerative disease of the central nervous system (CNS) characterized by demyelination and concomitant axonal loss. The lack of a single specific test, and the similarity to other inflammatory diseases of the central nervous system
Carolina Dalmasso et al.
Journal of the American Heart Association, 8(23), e012309-e012309 (2019-11-23)
Background We have previously reported that female mice exposed to maternal separation and early weaning (MSEW), a model of early life stress, show exacerbated diet-induced obesity associated with hypertension. The goal of this study was to test whether MSEW promotes
D Ostalska-Nowicka et al.
Journal of biological regulators and homeostatic agents, 33(2), 415-420 (2019-04-11)
Obesity-related glomerulopathy (ORG) is an increasingly detected syndrome present in children with obesity. Megalin, a constitutive proximal tubule cell protein, when present in urine, can be considered as a biomarker indicating renal injury in these children.
Chuanming Xu et al.
American journal of physiology. Renal physiology, 318(5), F1122-F1135 (2020-03-17)
Emerging evidence has demonstrated that (pro)renin receptor (PRR)-mediated activation of intrarenal renin-angiotensin system (RAS) plays an essential role in renal handling of Na+ and water balance and blood pressure. The present study tested the possibility that the intrarenal RAS served

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service