Skip to Content
Merck
All Photos(1)

Documents

RAB0980

Sigma-Aldrich

Phosphotyrosine MET / Hepatocyte Growth Factor Receptor ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

technique(s)

ELISA: suitable

assay range

inter-assay cv: <10%
intra-assay cv: <12%

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MET(4233)

General description

The antibody pair provided in this kit recognizes Human Tyrosine-Phosphorylated-MET.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABHRP6Phosphotyrosine ELISA HRP-StreptavidinSDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jesse Hudson et al.
Molecular and cellular biology, 34(19), 3689-3701 (2014-07-30)
Breast cancers are stratified into distinct subtypes, which influence therapeutic responsiveness and patient outcome. Patients with luminal breast cancers are often associated with a better prognosis relative to that with other subtypes. However, subsets of patients with luminal disease remain
Ji-Dong Chen et al.
Asian Pacific journal of cancer prevention : APJCP, 15(11), 4545-4548 (2014-06-28)
To investigate the changes of serum vascular endothelial growth factor (VEGF), soluble interleukin-2 receptor (SIL-2R) and hepatocyte growth factor (HGF) contents in patients with primary hepatocellular carcinoma (HCC) before and after percutaneous microwave coagulation therapy (PMCT) and determine their clinical
Mishaal B Patel et al.
Carcinogenesis, 35(8), 1891-1900 (2014-05-31)
Activated cancer-associated human pancreatic stellate cells (CAhPSCs, which produce the collagenous stroma of pancreatic cancer [PC]) are known to play a major role in PC progression. Apart from inducing cancer cell proliferation and migration, CAhPSCs have also been implicated in
Qian Wen et al.
Journal of translational medicine, 12, 114-114 (2014-06-03)
Autologous transplantation of modified mesenchymal stem cells (MSCs) is a promising candidate for the treatment of the refractory clinical disease, avascular necrosis of the femoral head (ANFH). Our previous attempts by compounding MSCs with medical fibrin glue to treat ANFH
Pei-Min Chen et al.
Journal of leukocyte biology, 96(2), 295-303 (2014-04-10)
Monocytes are a population of leukocytes that terminally differentiate into macrophages and DCs. Whereas these differentiated progeny have inflammatory and resident--which are more immunomodulatory--phenotypes, less has been reported on the plasticity of monocytes themselves. We found that MSCs, a population

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service