RAB0971
Phosphotyrosine JAK2 / Tyrosine-protein Kinase JAK2 ELISA Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
technique(s)
ELISA: suitable
assay range
inter-assay cv: <10%
intra-assay cv: <12%
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... JAK2(3717)
Related Categories
General description
The antibody pair provided in this kit recognizes Human Tyrosine-Phosphorylated-JAK2. The JAK2 (Janus kinase 2) gene is mapped to human chromosome 9p24.1.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
Janus kinase 2 (JAK2) mediates epithelial-mesenchymal transition in hepatocellular carcinoma. The JAK2/STAT3 signaling pathway is associated with tumor development in a number of cancer such as gastric cancer. Mutation in the JAK2 gene is known to activate JAK2 receptor action even in the absence of ligand, inducing cell proliferation and opposing apoptosis.
Kit Components Also Available Separately
Product No.
Description
SDS
- RABHRP6Phosphotyrosine ELISA HRP-StreptavidinSDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
CD 274 (PDL 1) and JAK 2 genomic amplifications in pulmonary squamous?cell and adenocarcinoma patients.
Clave S, et al.
Histopathology, 72(2), 259-269 (2018)
SIRT6 inhibits growth of gastric cancer by inhibiting JAK2/STAT3 pathway.
Zhou J, et al.
Oncology Reports, 38(2), 1059-1066 (2017)
Norcantharidin inhibits IL-6-induced epithelial?mesenchymal transition via the JAK2/STAT3/TWIST signaling pathway in hepatocellular carcinoma cells.
Gao Y, et al.
Oncology Reports, 38(2), 1224-1232 (2017)
The genetic basis of haematological cancers (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service