Skip to Content
Merck
All Photos(1)

Key Documents

RAB0739

Sigma-Aldrich

Human THBD / Thrombomodulin ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 8 pg/mL
standard curve range: 7.06-800 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... THBD(7056)

General description

The antibody pair provided in this kit recognizes Human Thrombomodulin.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph Kamtchum-Tatuene et al.
Neurology(R) neuroimmunology & neuroinflammation, 6(2), e531-e531 (2019-01-31)
To study the relationship between endothelial dysfunction, HIV infection, and stroke in Malawians. Using a cross-sectional design, we measured plasma levels of intercellular adhesion molecule-1 (ICAM-1), plasminogen activator inhibitor-1 (PAI-1), vascular endothelial growth factor (VEGF), and soluble thrombomodulin (sTM) in
Nadine Althof et al.
EMBO molecular medicine, 10(2), 200-218 (2018-01-04)
Severe heart pathology upon virus infection is closely associated with the immunological equipment of the host. Since there is no specific treatment available, current research focuses on identifying new drug targets to positively modulate predisposing immune factors. Utilizing a murine
Joshua Y C Yang et al.
Journal of clinical medicine, 8(2) (2019-02-20)
Standard methods for detecting chronic lung allograft dysfunction (CLAD) and rejection have poor sensitivity and specificity and have conventionally required bronchoscopies and biopsies. Plasma cell-free DNA (cfDNA) has been shown to be increased in various types of allograft injury in
K H Simons et al.
Journal of internal medicine, 282(6), 522-536 (2017-09-01)
Toll like receptors (TLR) play an important role in vein graft disease (VGD). Interferon regulatory factors (IRF) 3 and 7 are the transcriptional regulators of type I interferons (IFN) and type I IFN responsive genes and are downstream factors of
Mouna Habbane et al.
Genes, 11(9) (2020-09-02)
Leigh syndrome (LS) is a serious genetic disease that can be caused by mutations in dozens of different genes. Clinical study of a deafness pedigree in which some members developed LS. Cellular, biochemical and molecular genetic analyses of patients' tissues

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service