Skip to Content
Merck
All Photos(1)

Key Documents

RAB0674

Sigma-Aldrich

Human CGB / Choriogonadotropin Subunit Beta ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available.

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 40 pg/mL
standard curve range: 40.96-10000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CGB(1082)

General description

The antibody pair provided in this kit recognizes Human Chorionic Gonadotropin beta.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Chorionic gonadotropin β subunit 3 (CGB3), also referred as human chorionic gonadotropin- β (HCG-β) is a specific and active subunit, secreted by the syncytiotrophoblast of the placenta. It plays a vital role in embryo implantation.[1] In addition, it acts like an agonist of luteinizing hormone (LH), stimulating progesterone secretion by the corpus luteum.[2] Elevated expression of CGB3 enhance tumor invasion and is also used to predict poor prognosis of early-stage colorectal cancer (CRC).[1]

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Review: hCGs: different sources of production, different glycoforms and functions.
Fournier T, et al.
Placenta, 36(1), S60-S65 (2015)
B Subunit of Human Chorionic Gonadotropin Promotes Tumor Invasion and Predicts Poor Prognosis of Early-Stage Colorectal Cancer.
Li J, et al.
Cellular Physiology and Biochemistry, 45, 237-249 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service