Skip to Content
Merck
All Photos(2)

Key Documents

RAB0489

Sigma-Aldrich

Human soluble TNF Receptor 1 ELISA Kit

for serum, plasma, cell culture supernatants and urine

Synonym(s):

sTNFRI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84
Pricing and availability is not currently available.

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 pg/mL
standard curve range: 0.55-400 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TNFRSF1A(7132)

General description

The Human sTNF RI (soluble tumor necrosis factor receptor I) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme- linked immunosorbent assay for the quantitative measurement of human sTNF RI in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human TNF RI

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Tumour necrosis factor (TNF) receptor superfamily member 1A is a type I single transmembrane glycoprotein. Signal transduction mediated by binding of TNFα regulates inflammatory, immune responses and apoptotic pathway. TNF receptor 1 is implicated in the end-stage renal disease and associated mortality.[1] Mutations in TNF receptor 1 causes autosomal dominant tumor necrosis factor receptor-associated periodic syndrome (TRAPS).[2] Polymorphism in TNF receptor 1 causes multiple sclerosis.[3]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel TNFRSF1A gene mutation in a patient with tumor necrosis factor receptor-associated periodic syndrome
Khabazi A, et al.
Hematology/oncology and Stem Cell Therapy, 11(3), 175-177 (2018)
TNF receptor 1 genetic risk mirrors outcome of anti-TNF therapy in multiple sclerosis
Gregory AP, et al.
Nature, 488(7412), 508-511 (2012)
Circulating TNF receptors 1 and 2 predict mortality in patients with end-stage renal disease undergoing dialysis
Gohda T, et al.
Scientific Reports, 7, 43520-43520 (2017)
Kazuo Kubota et al.
Journal of clinical immunology, 34(5), 584-593 (2014-04-25)
To investigate clinical presentation, genetic background and cytokine profile of Japanese sporadic cases of periodic fever, aphthous stomatitis, pharyngitis and adenitis (PFAPA) syndrome. Nine PFAPA syndrome patients were recruited. DNA sequence analysis of auto inflammatory disorder susceptibility genes, MEFV, MVK
Anil K Taneja et al.
International journal of cardiology, 175(2), 253-260 (2014-06-01)
Heart failure activates neurohormones, and elevated levels of brain natriuretic peptide (BNP) are associated with adverse outcomes. The SENIORS trial showed that nebivolol, a highly selective beta-1 antagonist with vasodilating properties, reduced the composite outcome of all cause mortality or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service