RAB0456
Neurokinin-B EIA Kit
for serum, plasma, culture supernatant and cell lysates
About This Item
Recommended Products
species reactivity
mouse, rat, human
technique(s)
ELISA: suitable
enzyme immunoassay: suitable
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... TAC3(6866)
mouse ... TAC3(21334)
rat ... TAC3(29191)
Related Categories
General description
Immunogen
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service