Skip to Content
Merck
All Photos(1)

Key Documents

RAB0447

Sigma-Aldrich

Phospho-Stat3 (pTyr705) and pan-Stat3 ELISA Kit

for detection of phospho-stat3 (pTyr705) and pan-stat3 in human, mouse or rat cell and tissue lysates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available.

species reactivity

human, mouse, rat

technique(s)

ELISA: suitable
capture ELISA: suitable

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... STAT3(6774)
mouse ... STAT3(20848)
rat ... STAT3(25125)

General description

Signal transducers and activators of transcription 3 (STAT3) is a transcription factor that belongs to the family of STAT proteins. The protein contains an Src homology 2 (SH2) domain for protein-protein interactions.[1] STAT3 is activated by phosphorylation on tyrosine 705 by c-Jun N-terminal kinases (JNKs), cytokine receptor-associated kinases, nonreceptor tyrosine kinases, and growth factor receptor tyrosine kinases, in response to growth factors and cytokines.[1][2] Phosphorylation of STAT3 is important for dimer formation and gene expression.[1][2] STAT3 mediates the expression of a variety of genes that are involved in cell proliferation and differentiation. It also aids in pro-cancer immunity.[3] The Phospho-Stat3 (pTyr705) and pan-Stat3 ELISA (enzyme-linked immunosorbent assay) is a very rapid, convenient, and sensitive assay kit that can monitor the activation or function of important biological pathways in cell lysates.
The Phospho-Stat3 (pTyr705) ELISA (Enzyme-Linked Immunosorbent Assay) kit is a very rapid, convenient and sensitive assay kit that can monitor the activation or function of important biological pathways in cell lysates.

Immunogen

Stat3 (pTyr705) synthetic phosphopeptide, Recombinant Human Stat3

Application

Phospho-Stat3 (pTyr705) and pan-Stat3 ELISA Kit has been used in enzyme-linked immunosorbent assay (ELISA) for quantitative measurement of phosphorylated signal transducer and activator of transcription3 (p-STAT3) in liver tissues.[4][5]

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinbo Yang et al.
Cancer research, 65(3), 939-947 (2005-02-12)
Signal transducer and activator of transcription 3 (STAT3) is phosphorylated on tyrosine residue 705 in response to growth factors or cytokines to form activated homodimers that drive gene expression. Because the stat3 promoter has a binding site for STAT3 dimers
Targeting the Hepatocyte Growth Factor Receptor to Overcome Resistance to Targeted Therapies
Embryonic Stem Cell Protocols, 25-60 (2019)
Marisa Dolled-Filhart et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 9(2), 594-600 (2003-02-11)
Although a high frequency of tumors contain constitutively activated signal transducers and activators of transcription 3 (Stat3), its relationship to breast cancer and patient survival has not been determined in a large retrospective study of node-negative tumors. To further elucidate
Manar M Esmail et al.
Toxicology letters, 347, 23-35 (2021-05-08)
Liver fibrosis is the conjoint consequence of almost all chronic liver diseases. Cholestatic liver injury is a significant stimulus for fibrotic liver. This study was conducted to investigate the hepatoprotective effect of niclosamide as a NOTCH inhibitor and on the
Hyo Jeong Kang et al.
Oncotarget, 8(16), 27263-27276 (2017-05-04)
The non-receptor tyrosine phosphatase SHP2 has scaffolding functions in signal transduction cascades downstream of growth receptors. A recent study suggested that SHP2 acts as a tumor suppressor during hepatocellular carcinoma (HCC) development. Herein we examined whether SHP2 links the HBx-NF-κB

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service