Skip to Content
Merck
All Photos(1)

Documents

RAB0428

Sigma-Aldrich

Vaspin EIA Kit

for serum, plasma, culture supernatant and cell lysates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

species reactivity

human, rat, mouse

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

The Vaspin Enzyme Immunoassay (EIA) Kit is an in vitro quantitative assay for detecting Vaspin peptide based on the principle of Competitive Enzyme Immunoassay.

Immunogen

Vaspin, synthetic peptide

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vaspin as a Prognostic Marker in Patients with Acute Myocardial Infarction.
Zhang B
Heart, Lung & Circulation, 25, 257-264 (2016)
Hangyuan He et al.
Life sciences, 269, 119096-119096 (2021-01-23)
This study intends to explore the role of Vaspin and cholesterol metabolism in the process of osteoarthritis (OA) and its mechanism in vitro and in vivo. In vitro, chondrocytes were treated with interleukin-1β (IL-1β, 20 ng/mL) in combination with Vaspin at
Jan Pippel et al.
Biological chemistry, 397(2), 111-123 (2015-11-04)
The adipokine vaspin (serpinA12) is mainly expressed in white adipose tissue and exhibits various beneficial effects on obesity-related processes. Kallikrein 7 is the only known target protease of vaspin and is inhibited by the classical serpin inhibitory mechanism involving a
Association between vaspin level and coronary artery disease in patients with type 2 diabetes.
Hao F, et.al.
Diabetes Research and Clinical Practice, 113, 26-32 (2016)
Association between vaspin level and coronary artery disease in patients with type 2 diabetes.
Hao F
Diabetes Research and Clinical Practice, 113, 26-32 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service