RAB0368
Mouse MMP-3 ELISA Kit
for serum, plasma and cell culture supernatant
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.84
Recommended Products
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type serum
sample type cell culture supernatant(s)
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 13.72-10000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Mmp3(17392)
Related Categories
General description
The Mouse MMP-3 (Matrix metalloproteinase-3) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse MMP-3 in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Mouse MMP3
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tino Stauber et al.
Advanced healthcare materials, 10(20), e2100741-e2100741 (2021-09-09)
Tendons are among the most mechanically stressed tissues of the body, with a functional core of type-I collagen fibers maintained by embedded stromal fibroblasts known as tenocytes. The intrinsic load-bearing core compartment of tendon is surrounded, nourished, and repaired by
Faxin Li et al.
Inflammation, 37(4), 1134-1141 (2014-02-18)
Rheumatoid arthritis (RA) is a chronic autoimmune disease with high morbidity and mortality. Fibroblast-like synoviocytes (FLS) in the synovial tissues play critical roles in joint destruction. Recent studies implicate the sumoylation in the regulation of the inflammation and arthritis. Thus
Xue-Xuan Feng et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 63, 113-123 (2014-07-19)
Ultraviolet (UV) irradiation, known to generate reactive oxygen species (ROS) excessively and elicit inflammatory response, is a potent inducer for skin photoaging. Overproduction of ROS in conjunction with the resulting inflammation stimulate the over-expression of matrix metalloproteinases (MMPs), which in
C Andersson et al.
Clinical and experimental immunology, 177(1), 219-233 (2014-03-29)
Preclinical evidence supports targeting the C5a receptor (C5aR) in rheumatoid arthritis (RA). To support ongoing clinical development of an anti-C5aR monoclonal antibody, we have investigated for the first time the mechanism of action and the pharmacodynamics of a blocking anti-murine
Özge Erdemli et al.
Journal of biomaterials applications, 29(4), 524-542 (2014-05-24)
Anti-tumor necrosis factor α (TNFα) drugs such as etanercept (ETN) have been mostly used in systemic treatment of rheumatoid arthritis. To eliminate the side effects in long-term treatments and to achieve a local sustained anti-inflammatory effect, a controlled drug delivery
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service