RAB0335
Rat Leptin ELISA Kit
for serum, plasma and cell culture supernatant
Synonym(s):
Leptin, Obese protein, Obesity factor
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
rat
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type serum
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 30 pg/mL
standard curve range: 10.97-8000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
rat ... Lep(25608)
General description
The Rat Leptin ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of rat Leptin in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Rat Leptin
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
María Juárez-Fernández et al.
Antioxidants (Basel, Switzerland), 10(12) (2021-12-25)
Gut microbiota plays a key role in obesity and non-alcoholic fatty liver disease (NAFLD), so synbiotics could be a therapeutic alternative. We aim to evaluate a nutritional intervention together with the administration of the bacteria Akkermansia muciniphila and the antioxidant
József Szabó et al.
Acta veterinaria Hungarica, 69(2), 134-156 (2021-07-06)
The purpose of this 30-day feeding study was to elucidate the changes, correlations, and mechanisms caused by the replacement of the starch content of the AIN-93G diet (St) with glucose (G), fructose (F) or lard (L) in body and organ
Trevor Teich et al.
American journal of physiology. Endocrinology and metabolism, 311(1), E56-E68 (2016-05-05)
Severe caloric restriction (CR), in a setting of regular physical exercise, may be a stress that sets the stage for adiposity rebound and insulin resistance when the food restriction and exercise stop. In this study, we examined the effect of
Maria João Neuparth et al.
Journal of investigative medicine : the official publication of the American Federation for Clinical Research, 62(2), 350-353 (2013-11-13)
Physical exercise intervention is known to be crucial in the management of type 2 diabetes mellitus (T2DM). We aimed to evaluate, in patients with T2DM, the effect of regular moderate walking exercise on markers of oxidative stress, lipid metabolism, and
Simon G Anderson et al.
PloS one, 9(9), e103217-e103217 (2014-09-04)
Blood-vessel dysfunction arises before overt hyperglycemia in type-2 diabetes (T2DM). We hypothesised that a metabolomic approach might identify metabolites/pathways perturbed in this pre-hyperglycemic phase. To test this hypothesis and for specific metabolite hypothesis generation, serum metabolic profiling was performed in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service