RAB0333
Human Leptin ELISA Kit
for serum, plasma, cell culture supernatant and urine
Synonym(s):
Leptin, Obese protein, Obesity factor
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type urine
sample type plasma
sample type serum
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 1.64-400 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... LEP(3952)
General description
The Human Leptin ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human Leptin in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human Leptin
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
recommended
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chuang Peng et al.
International journal of oncology, 55(1), 298-308 (2019-05-23)
Leptin is an adipokine minimally known for its activities or underlying mechanisms in cholangiocarcinoma. The present study explored the effects of leptin on the epithelial‑mesenchymal transition (EMT) and pro‑angiogenic capability of cholangiocarcinoma cells, and investigated the underlying mechanisms. Cholangiocarcinoma cells were
Satoshi Sekiguchi et al.
Bioscience, biotechnology, and biochemistry, 75(4), 752-756 (2011-04-23)
Leptin is one of the representative adipocyte-derived protein hormones. Measuring the serum leptin concentration gives an important index for preventing and treating diabetes mellitus and other diseases. We constructed in this study a chemiluminescent enzyme immunoassay (CLEIA) for measuring leptin
Georgios K Dimitriadis et al.
Endocrine, 60(1), 175-184 (2018-02-06)
To explore the metabolic phenotype of obesity-related secondary hypogonadism (SH) in men pre-replacement and post-replacement therapy with long-acting intramuscular (IM) testosterone undecanoate (TU). A prospective observational pilot study on metabolic effects of TU IM in male obesity-related SH (hypogonadal [HG]
Shalini Behl et al.
Molecular biology reports, 46(3), 2643-2655 (2019-03-31)
Rilpivirine is a non-nucleoside reverse transcriptase inhibitor, recently developed as a drug of choice for initial anti-retroviral (ARV) treatment of HIV-1 infection, whereas estradiol is a major component of hormonal contraceptives. Both drugs have effects on lipid metabolism, impairment of
B Navia et al.
Acta physiologica (Oxford, England), 211(4), 585-596 (2014-06-18)
Interleukin-6 (IL-6) is a major cytokine controlling body weight and metabolism, but because many types of cells can synthesize and respond to IL-6 considerable uncertainty still exists about the mechanisms underlying IL-6 effects. Therefore, the aim of this study was
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service