Skip to Content
Merck
All Photos(2)

Documents

RAB0228

Sigma-Aldrich

Human IGF-I ELISA Kit

for serum, plasma, cell culture supernatant and urine

Synonym(s):

Insulin-like growth factor I, Mechano growth factor, Somatomedin-C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type urine
sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 100 pg/mL
standard curve range: 0.123-30 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IGF1(3479)

General description

The Human IGF-I ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IGF-I in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human IGF-I

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of insulin and insulin-like growth factors on cultured human hair follicles: IGF-I at physiologic concentrations is an important regulator of hair follicle growth in vitro.
Philpott MP, et. al.
The Journal of Investigative Dermatology, 102, 857-857 (1994)
Activation of the IGF1 pathway mediates changes in cellular contractility and motility in single-suture craniosynostosis.
Al-Rekabi Z
Journal of Cell Science, 129 (2016)
Various growth factor yields in various platelet rich plasma processing methods.
Sari P
BioTechnology: An Indian Journal, 9 (2014)
Effects of insulin and insulin-like growth factors on cultured human hair follicles: IGF-I at physiologic concentrations is an important regulator of hair follicle growth in vitro.
Philpott MP
The Journal of Investigative Dermatology, 102, 857-861 (1994)
Pablo Roman-Garcia et al.
The Journal of clinical investigation, 124(7), 2988-3002 (2014-06-10)
Both maternal and offspring-derived factors contribute to lifelong growth and bone mass accrual, although the specific role of maternal deficiencies in the growth and bone mass of offspring is poorly understood. In the present study, we have shown that vitamin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service