Skip to Content
Merck
All Photos(2)

Key Documents

RAB0116

Sigma-Aldrich

Human GRO-α / CXCL1 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available.

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type urine
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 1.37-1000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CXCL1(2919)

General description

The Human GRO-α (Growth Regulated Oncogene-α) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme- linked immunosorbent assay for the quantitative measurement of human GRO-α in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human GRO-α

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ziwei Zhou et al.
Frontiers in pharmacology, 13, 870178-870178 (2022-07-06)
The combination of immunotherapy with platinum-based chemotherapy has become the first-line treatment for patients with advanced non-small cell lung cancer (NSCLC) with negative driver gene mutations. However, finding an ideal chemotherapeutic regimen for immunotherapy and exploring the underlying mechanism have
Dominika Nackiewicz et al.
Nephron. Experimental nephrology, 126(3), 141-147 (2014-05-24)
Lipoprotein abnormalities are associated with a rapid decline in renal function in patients of chronic kidney disease. In addition, hyperlipidemia is associated with an increased risk of developing renal insufficiency. The underlying molecular mechanisms for these clinical findings are unclear.
Syed A Khurram et al.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology, 43(9), 667-674 (2014-06-27)
Chemokines regulate physiological and pathological leucocyte trafficking, and chemokine receptors play a role in tumorigenesis. Expression of interleukin-8 (IL-8) receptors CXCR1 and CXCR2 has been shown in oral squamous cell carcinoma (OSCC) but remains poorly characterised. This aim of this
Michael D Diem et al.
Protein engineering, design & selection : PEDS, 27(10), 419-429 (2014-05-03)
Alternative scaffold molecules represent a class of proteins important to the study of protein design and mechanisms of protein-protein interactions, as well as for the development of therapeutic proteins. Here, we describe the generation of a library built upon the
Elvira L Liclican et al.
Cancer prevention research (Philadelphia, Pa.), 7(8), 845-855 (2014-06-11)
Understanding the molecular pathogenesis of lung cancer is necessary to identify biomarkers/targets specific to individual airway molecular profiles and to identify options for targeted chemoprevention. Herein, we identify mechanisms by which loss of microRNA (miRNA)125a-3p (miR125a) contributes to the malignant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service