Skip to Content
Merck
All Photos(2)

Key Documents

RAB0051

Sigma-Aldrich

Human PARC / CCL18 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$693.00

$693.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
1 KIT
$693.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$693.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 2.74-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL18(6362)

General description

CC chemokine ligand 18 (CCL18) is expressed at high levels in human lung and serum. This CC chemokine can be found in primates only. The CCL18 gene is mapped to the human chromosome 17q11.2.[1]
The Human PARC ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human PARC in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human PARC

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

C-C motif chemokine ligand 18 (CCL18) has immunological functions and is associated with tumor progression. It is secreted by tumor associated macrophages (TAMs). The protein recruits naive T-cells and dendritic cells, and induces regulatory T-cells. It also acts as a chemokine for helper T lymphocytes and cytotoxic T lymphocytes. CCL18 has been implicated in the pathogenesis of many cancers.[2][3]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shinichiro Ohshimo et al.
Respiratory medicine, 108(7), 1031-1039 (2014-05-20)
Acute exacerbation (AE) is a major cause of death in idiopathic pulmonary fibrosis (IPF). However, little is known about sensitive biomarkers for predicting AE. The aim of our study was to investigate the significance of KL-6 and CC-Chemokine Ligand 18
25th European Photovoltaic Solar Energy Conference and Exhibition / 5th World Conference on Photovoltaic Energy Conversion; 6-10 September 2010; Valencia; Spain; EU PVSEC Proceedings null
CCL18 from tumor-cells promotes epithelial ovarian cancer metastasis via mTOR signaling pathway
Qi Wang
Molecular Carcinogenesis (2016)
Local Concentrations of CC-Chemokine-Ligand 18 Correlate with Tumor Size in Non-small Cell Lung Cancer and Are Elevated in Lymph Node-positive Disease.
Schmid S
Anticancer Research (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service