Skip to Content
Merck
All Photos(2)

Key Documents

RAB0029

Sigma-Aldrich

Human BMP-4 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available.

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type urine
sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 15 pg/mL
standard curve range: 8.23-6000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... BMP4(652)

General description

The Human BMP-4 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human BMP-4 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human BMP4

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABDIL6ELISA 5X Sample Diluent Buffer (Item D2)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simone Bersini et al.
eLife, 9 (2020-09-09)
Vascular dysfunctions are a common feature of multiple age-related diseases. However, modeling healthy and pathological aging of the human vasculature represents an unresolved experimental challenge. Here, we generated induced vascular endothelial cells (iVECs) and smooth muscle cells (iSMCs) by direct
Guillermo Agustín Videla Richardson et al.
Brain pathology (Zurich, Switzerland), 26(1), 43-61 (2015-03-27)
Although BMP4-induced differentiation of glioma stem cells (GSCs) is well recognized, details of the cellular responses triggered by this morphogen are still poorly defined. In this study, we established several GSC-enriched cell lines (GSC-ECLs) from high-grade gliomas. The expansion of
Yuan Cao et al.
Cancer research, 74(18), 5091-5102 (2014-09-17)
The TGFβ growth factor family member BMP4 is a potent suppressor of breast cancer metastasis. In the mouse, the development of highly metastatic mammary tumors is associated with an accumulation of myeloid-derived suppressor cells (MDSC), the numbers of which are
Silvia Calpe et al.
Molecular cancer therapeutics, 14(11), 2527-2540 (2015-09-10)
Bone morphogenetic proteins (BMP) have important but distinct roles in tissue homeostasis and disease, including carcinogenesis and tumor progression. A large number of BMP inhibitors are available to study BMP function; however, as most of these antagonists are promiscuous, evaluating

Questions

  1. Can you please provide information on the detection of amino acids, including the specific range?

    1 answer
    1. We have not conducted epitope mapping, but the available antibody specificity information is provided below:

      Capture and Detection Antibody:
      - Source: Monoclonal Mouse IgG2B
      - Immunogen: Mouse myeloma cell line NS0-derived recombinant human BMP-4 (aa293-408)
      - Accession# P12644

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service