Key Documents
RA481WO-EP
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
A-4-81, for 5810/5810R, aerosol-tight, without buckets
About This Item
UNSPSC Code:
41103909
Pricing and availability is not currently available.
feature
to hold 4 x 500 mL rectangular buckets
to hold 4 x MTP/flex buckets
aerosol-tight
for 5810/5810R
medium to high-capacity swing-bucket rotor
without buckets
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022638611 (NA)
Eppendorf® 5810743001 (EU)
parameter
4,000 max. RCF (3,220 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
Related Categories
General description
A wide selections of rotors to fit the Eppendorf 5804 and 5810 centrifuges.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)