Skip to Content
Merck
All Photos(1)

Key Documents

R9388

Sigma-Aldrich

Retinol Binding Protein from human urine

buffered aqueous solution

Synonym(s):

CRBP, Cellular retinol binding protein, RBP, RBP-1, Retinol Binding Protein I

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56
Pricing and availability is not currently available.

biological source

human urine

Quality Level

Assay

≥85% (SDS-GE)

form

buffered aqueous solution

technique(s)

cell culture | embryo: suitable
transfection: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... RBP1(5947)

General description

Retinol Binding Protein (RBP) is a member of lipocalin superfamily.[1] The normal concentration of RBP in serum is 40 to 50 μg/mL and has a molecular weight of 21 kDa. It has a turnover rate of 4 to 24 h.[2] RBP is produced in the liver in response to retinol binding trigger. It is also produced in kidney, sertoli cells of testis.[1] RBP contains a single globular domain and a highly conserved N-terminal region. It has eight antiparallel β sheets in the centre which binds retinol.[3] In human chromosome, the gene encoding RBP is localised at 3q23.[4]

Application

Retinol Binding Protein from human urine has been used:
  • as one of the protein in microinjecting mesencephalic cavity of chicken eggs to monitor embryonic cerebrospinal fluid/embryonic serum protein transfer[5]
  • as an additive in basal medium for RNA transfection experiments[6]
  • in the antibody-derivatized affinity pipettes preparation for the capture of affinity proteins[7]

Biochem/physiol Actions

Inflammation induced by lipopolysaccharide (LPS) exposure caused a reduction in the level of retinol binding protein produced by the liver. [8]
Retinol Binding Protein (RBP) is a specific protein for the transport of vitamin A. The apo-RBP, formed after delivering retinol to the tissues is catabolized by the kidney. The plasma concentration of RBP is less in diabetes mellitus patients.[2] RBP binds to the carrier protein transerythrin, to prevent its loss during glomerular filtration.[1] RBP is implicated in initial neurogenesis and neuroepithelial cell proliferation.[5] RBP is responsible for blepharophimosis syndrome (BPES).[4]

Physical form

Solution in 10 mM sodium phosphate, pH 7.4, 150 mM NaCl, and 0.05% NaN3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assignment of the cellular retinol-binding protein 1 gene (RBP1) and of the coatomer beta' subunit gene (COPB2) to human chromosome band 3q23 by in situ hybridization
De Baere E, et al.
Cytogenetics and Cell Genetics, 82, 226-227 (1998)
F J Rosales et al.
Journal of lipid research, 37(5), 962-971 (1996-05-01)
The acute inflammatory response to tissue injury and infection is associated with low concentrations of plasma retinol and its specific transport proteins, retinol-binding protein (RBP) and transthyretin (TTR). To examine the kinetics and mechanism of hyporetinemia, we have induced acute
Vitamin A in Health and Disease, 87-95 (1994)
Plasma retinol binding protein: structure and function of the prototypic lipocalin
Newcomer ME and Ong DE
Biochimica et Biophysica Acta, Protein Structure and Molecular Enzymology, 1482(1-2), 57-64 (2000)
A blood-CSF barrier function controls embryonic CSF protein composition and homeostasis during early CNS development
Parvas M, et al.
Developmental Biology, 321(1), 51-63 (2008)

Questions

  1. Which protein is this? RBP4?

    1 answer
    1. Retinol Binding Protein is a transport protein of Vitamin A. There are a number of publications available regarding the biological activtity and functionality of this protein. See the link below to access a review article that may be of interest:
      https://www.frontiersin.org/articles/10.3389/fphys.2021.659977/full

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service